Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101582698-101582957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLL
Entrez Gene ID
27343
Gene Name
polymerase (DNA) lambda
Gene Aliases
BETAN, POLKAPPA
UniGene
Hs.523230
Gene Chromosome Location
Chr.10: 101578882-101588270 on Build GRCh38
Transcripts
NM_001174084.1 NM_001174085.1 NM_001308382.1 NM_013274.3 NR_033406.1 XM_006717775.1 XM_006717776.1 XM_006717777.1 XM_011539650.1 XM_011539651.1 XM_011539652.1 XM_011539653.1 XM_011539654.1 XM_011539655.1 XM_011539656.1 XM_011539657.1 XM_011539659.1 XM_011539660.1 XM_011539662.1 XM_011539663.1 XM_011539664.1 XM_011539665.2 XM_011539666.1 XM_011539667.1 XM_017016084.1 XM_017016085.1 XM_017016086.1 XM_017016087.1 XM_017016088.1 XM_017016089.1 XM_017016090.1 XM_017016091.1 XM_017016092.1 XM_017016093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6366687 c.1193C>T p.T398I Chr.10: 101582764-101582764 on Build GRCh38
4011133 c.1127G>A p.G376D Chr.10: 101582830-101582830 on Build GRCh38
5433683 c.807_809delAGA p.E269delE Chr.10: 101582872-101582874 on Build GRCh38
1702936 c.1162C>T p.P388S Chr.10: 101582795-101582795 on Build GRCh38
4173344 c.846C>A p.A282A Chr.10: 101582835-101582835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55727130 Chr.10: 101582885-101582885 on Build GRCh38
rs1044610321 Chr.10: 101582769-101582769 on Build GRCh38
rs963299430 Chr.10: 101582870-101582870 on Build GRCh38
rs370590358 Chr.10: 101582859-101582859 on Build GRCh38
rs771498558 Chr.10: 101582721-101582721 on Build GRCh38
rs578000514 Chr.10: 101582748-101582748 on Build GRCh38
rs752786316 Chr.10: 101582767-101582768 on Build GRCh38
rs150900080 Chr.10: 101582798-101582798 on Build GRCh38
rs760315715 Chr.10: 101582739-101582739 on Build GRCh38
rs767305520 Chr.10: 101582762-101582762 on Build GRCh38
rs770606182 Chr.10: 101582746-101582746 on Build GRCh38
rs752071834 Chr.10: 101582890-101582890 on Build GRCh38
rs989248303 Chr.10: 101582795-101582795 on Build GRCh38
rs141596425 Chr.10: 101582766-101582766 on Build GRCh38
rs200623399 Chr.10: 101582831-101582831 on Build GRCh38
rs41291484 Chr.10: 101582832-101582832 on Build GRCh38
rs575920065 Chr.10: 101582857-101582857 on Build GRCh38
rs561184346 Chr.10: 101582904-101582904 on Build GRCh38
rs545400809 Chr.10: 101582801-101582801 on Build GRCh38
rs758574503 Chr.10: 101582915-101582915 on Build GRCh38
rs764077761 Chr.10: 101582909-101582909 on Build GRCh38
rs745469904 Chr.10: 101582896-101582896 on Build GRCh38
rs772907815 Chr.10: 101582772-101582772 on Build GRCh38
rs139871590 Chr.10: 101582891-101582891 on Build GRCh38
rs369790337 Chr.10: 101582800-101582800 on Build GRCh38
rs755508286 Chr.10: 101582865-101582865 on Build GRCh38
rs774342983 Chr.10: 101582928-101582928 on Build GRCh38
rs373311103 Chr.10: 101582910-101582910 on Build GRCh38
rs149619701 Chr.10: 101582856-101582856 on Build GRCh38
rs769587520 Chr.10: 101582897-101582897 on Build GRCh38
rs573710641 Chr.10: 101582822-101582822 on Build GRCh38
rs749818905 Chr.10: 101582863-101582863 on Build GRCh38
rs377248258 Chr.10: 101582731-101582731 on Build GRCh38
rs775197026 Chr.10: 101582902-101582902 on Build GRCh38
rs759779935 Chr.10: 101582780-101582780 on Build GRCh38
rs146112511 Chr.10: 101582866-101582866 on Build GRCh38
rs373429626 Chr.10: 101582829-101582829 on Build GRCh38

View Full Product Details