Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 55343363-55343622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR4A16
Entrez Gene ID
81327
Gene Name
olfactory receptor family 4 subfamily A member 16
Gene Aliases
OR11-117, OR4A16Q
UniGene
Hs.554530
Gene Chromosome Location
Chr.11: 55343201-55344187 on Build GRCh38
Transcripts
NM_001005274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133198 c.411_412GG>CT p.V138F Chr.11: 55343611-55343612 on Build GRCh38
3448780 c.406C>T p.R136* Chr.11: 55343606-55343606 on Build GRCh38
4433778 c.325G>A p.E109K Chr.11: 55343525-55343525 on Build GRCh38
6133200 c.347T>A p.M116K Chr.11: 55343547-55343547 on Build GRCh38
3986229 c.302T>C p.I101T Chr.11: 55343502-55343502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754051617 Chr.11: 55343557-55343557 on Build GRCh38
rs145395600 Chr.11: 55343566-55343566 on Build GRCh38
rs751496770 Chr.11: 55343599-55343599 on Build GRCh38
rs752331417 Chr.11: 55343418-55343418 on Build GRCh38
rs561603770 Chr.11: 55343587-55343587 on Build GRCh38
rs267602961 Chr.11: 55343593-55343593 on Build GRCh38
rs775163959 Chr.11: 55343456-55343456 on Build GRCh38
rs764187150 Chr.11: 55343598-55343598 on Build GRCh38
rs373385970 Chr.11: 55343500-55343500 on Build GRCh38
rs575051490 Chr.11: 55343558-55343558 on Build GRCh38
rs116541488 Chr.11: 55343513-55343513 on Build GRCh38
rs769408640 Chr.11: 55343579-55343579 on Build GRCh38
rs755507468 Chr.11: 55343487-55343487 on Build GRCh38
rs187469362 Chr.11: 55343389-55343389 on Build GRCh38
rs967014232 Chr.11: 55343510-55343510 on Build GRCh38
rs767151440 Chr.11: 55343416-55343416 on Build GRCh38
rs1011091844 Chr.11: 55343505-55343505 on Build GRCh38
rs202043396 Chr.11: 55343490-55343490 on Build GRCh38
rs761397240 Chr.11: 55343486-55343486 on Build GRCh38
rs774499821 Chr.11: 55343411-55343411 on Build GRCh38
rs780587336 Chr.11: 55343399-55343399 on Build GRCh38
rs74777917 Chr.11: 55343537-55343537 on Build GRCh38
rs369769854 Chr.11: 55343446-55343446 on Build GRCh38
rs755910896 Chr.11: 55343575-55343576 on Build GRCh38
rs775629849 Chr.11: 55343458-55343459 on Build GRCh38
rs577302078 Chr.11: 55343392-55343392 on Build GRCh38
rs755822753 Chr.11: 55343420-55343420 on Build GRCh38
rs142253044 Chr.11: 55343458-55343458 on Build GRCh38
rs374770148 Chr.11: 55343495-55343495 on Build GRCh38
rs372615022 Chr.11: 55343470-55343470 on Build GRCh38
rs200261293 Chr.11: 55343401-55343401 on Build GRCh38
rs148621575 Chr.11: 55343431-55343431 on Build GRCh38
rs115773757 Chr.11: 55343533-55343533 on Build GRCh38
rs375117618 Chr.11: 55343406-55343406 on Build GRCh38
rs750255280 Chr.11: 55343508-55343511 on Build GRCh38
rs757204611 Chr.11: 55343409-55343410 on Build GRCh38
rs147641414 Chr.11: 55343442-55343442 on Build GRCh38
rs764086893 Chr.11: 55343555-55343555 on Build GRCh38
rs765397400 Chr.11: 55343542-55343542 on Build GRCh38
rs142852989 Chr.11: 55343454-55343454 on Build GRCh38
rs781251889 Chr.11: 55343440-55343440 on Build GRCh38
rs767551999 Chr.11: 55343469-55343469 on Build GRCh38
rs150328499 Chr.11: 55343452-55343452 on Build GRCh38
rs201952971 Chr.11: 55343434-55343434 on Build GRCh38
rs778046694 Chr.11: 55343508-55343508 on Build GRCh38
rs752437404 Chr.11: 55343472-55343472 on Build GRCh38
rs553294477 Chr.11: 55343551-55343551 on Build GRCh38
rs902795310 Chr.11: 55343435-55343435 on Build GRCh38
rs371963500 Chr.11: 55343415-55343415 on Build GRCh38
rs765468983 Chr.11: 55343423-55343423 on Build GRCh38
rs868704666 Chr.11: 55343527-55343527 on Build GRCh38
rs373954394 Chr.11: 55343405-55343405 on Build GRCh38
rs139070294 Chr.11: 55343476-55343476 on Build GRCh38
rs149142469 Chr.11: 55343567-55343567 on Build GRCh38
rs749486335 Chr.11: 55343509-55343509 on Build GRCh38
rs767790290 Chr.11: 55343584-55343584 on Build GRCh38
rs76072904 Chr.11: 55343595-55343595 on Build GRCh38
rs145913777 Chr.11: 55343549-55343549 on Build GRCh38
rs76425968 Chr.11: 55343596-55343596 on Build GRCh38
rs763768417 Chr.11: 55343464-55343464 on Build GRCh38
rs144905099 Chr.11: 55343580-55343580 on Build GRCh38
rs761456259 Chr.11: 55343539-55343539 on Build GRCh38
rs543501126 Chr.11: 55343404-55343404 on Build GRCh38
rs775327695 Chr.11: 55343525-55343525 on Build GRCh38
rs373691578 Chr.11: 55343583-55343583 on Build GRCh38
rs768789430 Chr.11: 55343403-55343403 on Build GRCh38
rs758332856 Chr.11: 55343564-55343564 on Build GRCh38
rs544652707 Chr.11: 55343571-55343571 on Build GRCh38
rs779328620 Chr.11: 55343559-55343559 on Build GRCh38
rs747747041 Chr.11: 55343575-55343575 on Build GRCh38
rs374379880 Chr.11: 55343562-55343573 on Build GRCh38
rs116924453 Chr.11: 55343427-55343427 on Build GRCh38
rs368818886 Chr.11: 55343498-55343498 on Build GRCh38
rs142044895 Chr.11: 55343437-55343437 on Build GRCh38
rs762002982 Chr.11: 55343540-55343540 on Build GRCh38
rs751940185 Chr.11: 55343543-55343543 on Build GRCh38
rs76791457 Chr.11: 55343581-55343581 on Build GRCh38
rs532040362 Chr.11: 55343441-55343441 on Build GRCh38
rs147217176 Chr.11: 55343402-55343402 on Build GRCh38
rs79540892 Chr.11: 55343569-55343569 on Build GRCh38
rs745752609 Chr.11: 55343518-55343518 on Build GRCh38
rs772112660 Chr.11: 55343523-55343523 on Build GRCh38
rs777900222 Chr.11: 55343445-55343445 on Build GRCh38
rs780533927 Chr.11: 55343430-55343430 on Build GRCh38
rs368501875 Chr.11: 55343410-55343410 on Build GRCh38
rs376483129 Chr.11: 55343512-55343512 on Build GRCh38
rs147336887 Chr.11: 55343465-55343465 on Build GRCh38

View Full Product Details