Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17516731-17516990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51AP2
Entrez Gene ID
729475
Gene Name
RAD51 associated protein 2
Gene Aliases
-
UniGene
Hs.515802
Gene Chromosome Location
Chr.2: 17510584-17530414 on Build GRCh38
Transcripts
NM_001099218.2 NM_001321233.1 XM_011533084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3837460 c.1640T>A p.I547N Chr.2: 17516776-17516776 on Build GRCh38
7214000 c.1443T>A p.G481G Chr.2: 17516973-17516973 on Build GRCh38
4423330 c.1619_1620insA p.Q541fs*31 Chr.2: 17516796-17516797 on Build GRCh38
7213984 c.1442G>T p.G481V Chr.2: 17516974-17516974 on Build GRCh38
4467902 c.1513C>T p.P505S Chr.2: 17516903-17516903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756315257 Chr.2: 17516944-17516944 on Build GRCh38
rs62130402 Chr.2: 17516818-17516818 on Build GRCh38
rs752346694 Chr.2: 17516871-17516871 on Build GRCh38
rs763885940 Chr.2: 17516911-17516911 on Build GRCh38
rs988814789 Chr.2: 17516866-17516866 on Build GRCh38
rs368638717 Chr.2: 17516964-17516964 on Build GRCh38
rs763383317 Chr.2: 17516832-17516832 on Build GRCh38
rs781514390 Chr.2: 17516808-17516808 on Build GRCh38
rs759088226 Chr.2: 17516840-17516840 on Build GRCh38
rs375739867 Chr.2: 17516939-17516939 on Build GRCh38
rs370567024 Chr.2: 17516782-17516782 on Build GRCh38
rs755131551 Chr.2: 17516950-17516950 on Build GRCh38
rs747147361 Chr.2: 17516794-17516794 on Build GRCh38
rs757249553 Chr.2: 17516935-17516935 on Build GRCh38
rs780228921 Chr.2: 17516793-17516793 on Build GRCh38
rs576390249 Chr.2: 17516829-17516829 on Build GRCh38
rs752405616 Chr.2: 17516779-17516779 on Build GRCh38
rs757269788 Chr.2: 17516792-17516792 on Build GRCh38
rs773835144 Chr.2: 17516835-17516835 on Build GRCh38
rs767056429 Chr.2: 17516852-17516852 on Build GRCh38
rs374826217 Chr.2: 17516802-17516802 on Build GRCh38
rs753787790 Chr.2: 17516789-17516789 on Build GRCh38
rs62130403 Chr.2: 17516824-17516824 on Build GRCh38
rs767256329 Chr.2: 17516774-17516774 on Build GRCh38
rs974286251 Chr.2: 17516961-17516961 on Build GRCh38
rs773686203 Chr.2: 17516883-17516883 on Build GRCh38
rs865925575 Chr.2: 17516960-17516960 on Build GRCh38
rs934540093 Chr.2: 17516841-17516841 on Build GRCh38
rs771439086 Chr.2: 17516833-17516833 on Build GRCh38
rs748341414 Chr.2: 17516893-17516893 on Build GRCh38
rs199552327 Chr.2: 17516934-17516934 on Build GRCh38
rs376615884 Chr.2: 17516954-17516954 on Build GRCh38
rs748436615 Chr.2: 17516816-17516816 on Build GRCh38
rs760277182 Chr.2: 17516884-17516884 on Build GRCh38
rs1057018240 Chr.2: 17516853-17516853 on Build GRCh38
rs576895130 Chr.2: 17516814-17516814 on Build GRCh38
rs62130404 Chr.2: 17516946-17516946 on Build GRCh38
rs748358072 Chr.2: 17516959-17516959 on Build GRCh38
rs62130405 Chr.2: 17516953-17516953 on Build GRCh38

View Full Product Details