Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41877949-41878188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREBF2
Entrez Gene ID
6721
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, bHLHd2
UniGene
Hs.443258
Gene Chromosome Location
Chr.22: 41833079-41907308 on Build GRCh38
Transcripts
NM_004599.3 NR_103834.1 XM_006724310.2 XM_011530347.1 XM_017028921.1 XM_017028922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410967 c.1637G>T p.G546V Chr.22: 41877999-41877999 on Build GRCh38
1318761 c.1660G>A p.V554M Chr.22: 41878022-41878022 on Build GRCh38
3554996 c.1731C>T p.H577H Chr.22: 41878093-41878093 on Build GRCh38
5981859 c.1597G>A p.D533N Chr.22: 41877959-41877959 on Build GRCh38
5683240 c.1592G>C p.W531S Chr.22: 41877954-41877954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764418895 Chr.22: 41877971-41877971 on Build GRCh38
rs17002714 Chr.22: 41877968-41877968 on Build GRCh38
rs762284189 Chr.22: 41878069-41878069 on Build GRCh38
rs557989374 Chr.22: 41878066-41878066 on Build GRCh38
rs760750702 Chr.22: 41878031-41878031 on Build GRCh38
rs139907990 Chr.22: 41878096-41878096 on Build GRCh38
rs770776535 Chr.22: 41878122-41878122 on Build GRCh38
rs753296874 Chr.22: 41877998-41877999 on Build GRCh38
rs772165330 Chr.22: 41878136-41878136 on Build GRCh38
rs761130824 Chr.22: 41878065-41878065 on Build GRCh38
rs9623465 Chr.22: 41878030-41878030 on Build GRCh38
rs199937728 Chr.22: 41878072-41878072 on Build GRCh38
rs754511598 Chr.22: 41878000-41878000 on Build GRCh38
rs760831137 Chr.22: 41878139-41878139 on Build GRCh38
rs149832406 Chr.22: 41878131-41878131 on Build GRCh38
rs759659531 Chr.22: 41878055-41878055 on Build GRCh38
rs751057996 Chr.22: 41877994-41877994 on Build GRCh38
rs769230204 Chr.22: 41878142-41878142 on Build GRCh38
rs781174022 Chr.22: 41878009-41878009 on Build GRCh38
rs772103833 Chr.22: 41878063-41878063 on Build GRCh38
rs750070440 Chr.22: 41877974-41877974 on Build GRCh38
rs777697017 Chr.22: 41878118-41878118 on Build GRCh38
rs754206086 Chr.22: 41878068-41878068 on Build GRCh38
rs375617199 Chr.22: 41878117-41878117 on Build GRCh38
rs751227141 Chr.22: 41878095-41878095 on Build GRCh38
rs199860238 Chr.22: 41877993-41877993 on Build GRCh38
rs769599407 Chr.22: 41878016-41878016 on Build GRCh38
rs766063361 Chr.22: 41878074-41878074 on Build GRCh38
rs780606776 Chr.22: 41878101-41878101 on Build GRCh38
rs866787145 Chr.22: 41877980-41877980 on Build GRCh38
rs779096413 Chr.22: 41878128-41878128 on Build GRCh38
rs749012797 Chr.22: 41878121-41878121 on Build GRCh38
rs112193984 Chr.22: 41878125-41878125 on Build GRCh38
rs777500819 Chr.22: 41878020-41878020 on Build GRCh38
rs922076827 Chr.22: 41878086-41878086 on Build GRCh38
rs777283664 Chr.22: 41878148-41878148 on Build GRCh38
rs774557555 Chr.22: 41878054-41878054 on Build GRCh38
rs935634912 Chr.22: 41878152-41878152 on Build GRCh38
rs752281678 Chr.22: 41878112-41878112 on Build GRCh38
rs374060553 Chr.22: 41877982-41877982 on Build GRCh38
rs747947350 Chr.22: 41878067-41878067 on Build GRCh38
rs768690705 Chr.22: 41878138-41878138 on Build GRCh38
rs748085134 Chr.22: 41878015-41878015 on Build GRCh38
rs546034262 Chr.22: 41878064-41878064 on Build GRCh38

View Full Product Details