Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 43796997-43797262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM117
Entrez Gene ID
84216
Gene Name
transmembrane protein 117
Gene Aliases
-
UniGene
Hs.444668
Gene Chromosome Location
Chr.12: 43795806-44389762 on Build GRCh38
Transcripts
XM_011538831.2 NM_001242397.1 NM_002822.4 NR_073472.1 NR_073473.1

Gene Symbol
TWF1
Entrez Gene ID
5756
Gene Name
twinfilin actin binding protein 1
Gene Aliases
A6, PTK9
UniGene
Hs.189075
Gene Chromosome Location
Chr.12: 43793723-43806375 on Build GRCh38
Transcripts
XM_011538831.2 NM_001242397.1 NM_002822.4 NR_073472.1 NR_073473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4874849 c.832C>A p.L278M Chr.12: 43797047-43797047 on Build GRCh38
6000717 c.827G>A p.R276Q Chr.12: 43797052-43797052 on Build GRCh38
6403415 c.829A>T p.M277L Chr.12: 43797050-43797050 on Build GRCh38
4949106 c.866T>G p.I289S Chr.12: 43797013-43797013 on Build GRCh38
6264050 c.849G>A p.K283K Chr.12: 43797030-43797030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947862217 Chr.12: 43797189-43797189 on Build GRCh38
rs113477916 Chr.12: 43797158-43797158 on Build GRCh38
rs746444085 Chr.12: 43797052-43797052 on Build GRCh38
rs989396173 Chr.12: 43797154-43797154 on Build GRCh38
rs770428704 Chr.12: 43797053-43797053 on Build GRCh38
rs757940609 Chr.12: 43797034-43797034 on Build GRCh38
rs772109830 Chr.12: 43797141-43797163 on Build GRCh38
rs764948734 Chr.12: 43797030-43797030 on Build GRCh38
rs112536702 Chr.12: 43797163-43797163 on Build GRCh38
rs527466125 Chr.12: 43797105-43797105 on Build GRCh38
rs775654853 Chr.12: 43797065-43797065 on Build GRCh38
rs767653842 Chr.12: 43797120-43797120 on Build GRCh38
rs756874040 Chr.12: 43797048-43797048 on Build GRCh38
rs759415044 Chr.12: 43797125-43797125 on Build GRCh38
rs774353461 Chr.12: 43797059-43797060 on Build GRCh38
rs61933174 Chr.12: 43797228-43797228 on Build GRCh38
rs888134598 Chr.12: 43797184-43797184 on Build GRCh38
rs745934873 Chr.12: 43797115-43797117 on Build GRCh38
rs765243009 Chr.12: 43797127-43797127 on Build GRCh38
rs370636775 Chr.12: 43797026-43797026 on Build GRCh38
rs547099125 Chr.12: 43797117-43797117 on Build GRCh38
rs752549476 Chr.12: 43797032-43797032 on Build GRCh38
rs777514139 Chr.12: 43797040-43797040 on Build GRCh38
rs768716931 Chr.12: 43797083-43797083 on Build GRCh38
rs752395489 Chr.12: 43797132-43797132 on Build GRCh38
rs749682782 Chr.12: 43797076-43797076 on Build GRCh38
rs916516954 Chr.12: 43797162-43797162 on Build GRCh38
rs147143603 Chr.12: 43797051-43797051 on Build GRCh38
rs763634711 Chr.12: 43797140-43797140 on Build GRCh38
rs112345200 Chr.12: 43797071-43797071 on Build GRCh38
rs773288493 Chr.12: 43797121-43797121 on Build GRCh38
rs758255873 Chr.12: 43797136-43797136 on Build GRCh38
rs1046304129 Chr.12: 43797212-43797212 on Build GRCh38
rs374701874 Chr.12: 43797043-43797043 on Build GRCh38
rs571621512 Chr.12: 43797142-43797142 on Build GRCh38

View Full Product Details