Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68537574-68537823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B17
Entrez Gene ID
7367
Gene Name
UDP glucuronosyltransferase family 2 member B17
Gene Aliases
BMND12, UDPGT2B17
UniGene
Hs.575083
Gene Chromosome Location
Chr.4: 68537184-68568527 on Build GRCh38
Transcripts
NM_001077.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546537 c.1417G>A p.G473R Chr.4: 68537801-68537801 on Build GRCh38
5549983 c.1438G>A p.A480T Chr.4: 68537780-68537780 on Build GRCh38
5667678 c.1464G>A p.Q488Q Chr.4: 68537754-68537754 on Build GRCh38
3669292 c.1568C>A p.T523K Chr.4: 68537650-68537650 on Build GRCh38
6816404 c.1437C>T p.V479V Chr.4: 68537781-68537781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766359615 Chr.4: 68537774-68537774 on Build GRCh38
rs903105152 Chr.4: 68537736-68537736 on Build GRCh38
rs370815401 Chr.4: 68537650-68537650 on Build GRCh38
rs749182572 Chr.4: 68537633-68537633 on Build GRCh38
rs774103180 Chr.4: 68537727-68537727 on Build GRCh38
rs148336920 Chr.4: 68537797-68537797 on Build GRCh38
rs998752399 Chr.4: 68537760-68537760 on Build GRCh38
rs112076153 Chr.4: 68537638-68537638 on Build GRCh38
rs760618760 Chr.4: 68537757-68537757 on Build GRCh38
rs558443905 Chr.4: 68537666-68537666 on Build GRCh38
rs74896325 Chr.4: 68537676-68537676 on Build GRCh38
rs748876551 Chr.4: 68537715-68537715 on Build GRCh38
rs539157531 Chr.4: 68537598-68537598 on Build GRCh38
rs780696179 Chr.4: 68537714-68537714 on Build GRCh38
rs750396059 Chr.4: 68537630-68537630 on Build GRCh38
rs758457306 Chr.4: 68537661-68537661 on Build GRCh38
rs368302186 Chr.4: 68537747-68537747 on Build GRCh38
rs770369463 Chr.4: 68537686-68537686 on Build GRCh38
rs377616205 Chr.4: 68537705-68537705 on Build GRCh38
rs145103523 Chr.4: 68537754-68537754 on Build GRCh38
rs374496520 Chr.4: 68537784-68537784 on Build GRCh38
rs764133190 Chr.4: 68537786-68537786 on Build GRCh38
rs757685627 Chr.4: 68537679-68537679 on Build GRCh38
rs375288266 Chr.4: 68537683-68537683 on Build GRCh38
rs1043038830 Chr.4: 68537682-68537682 on Build GRCh38
rs186138322 Chr.4: 68537780-68537780 on Build GRCh38
rs377546716 Chr.4: 68537631-68537631 on Build GRCh38
rs763057562 Chr.4: 68537785-68537785 on Build GRCh38
rs768251747 Chr.4: 68537724-68537724 on Build GRCh38
rs762710938 Chr.4: 68537599-68537599 on Build GRCh38
rs767195299 Chr.4: 68537622-68537622 on Build GRCh38
rs761323185 Chr.4: 68537739-68537739 on Build GRCh38
rs371257307 Chr.4: 68537775-68537775 on Build GRCh38
rs778033250 Chr.4: 68537665-68537665 on Build GRCh38
rs746393909 Chr.4: 68537685-68537685 on Build GRCh38
rs774143630 Chr.4: 68537617-68537617 on Build GRCh38
rs375055477 Chr.4: 68537606-68537606 on Build GRCh38
rs761708963 Chr.4: 68537619-68537619 on Build GRCh38
rs141367736 Chr.4: 68537640-68537640 on Build GRCh38

View Full Product Details