Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33500189-33500462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAK16
Entrez Gene ID
84549
Gene Name
MAK16 homolog
Gene Aliases
MAK16L, RBM13
UniGene
Hs.583805
Gene Chromosome Location
Chr.8: 33485167-33501260 on Build GRCh38
Transcripts
NM_032509.3 NM_001102401.2 NM_001265581.1 NM_025115.3 XM_011544665.2

Gene Symbol
TTI2
Entrez Gene ID
80185
Gene Name
TELO2 interacting protein 2
Gene Aliases
C8orf41, MRT39
UniGene
Hs.742048
Gene Chromosome Location
Chr.8: 33498509-33513601 on Build GRCh38
Transcripts
NM_032509.3 NM_001102401.2 NM_001265581.1 NM_025115.3 XM_011544665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246985 c.1414C>G p.R472G Chr.8: 33500336-33500336 on Build GRCh38
4426548 c.1406C>A p.S469Y Chr.8: 33500344-33500344 on Build GRCh38
5874608 c.1400G>A p.R467H Chr.8: 33500350-33500350 on Build GRCh38
4938621 c.1345T>C p.S449P Chr.8: 33500405-33500405 on Build GRCh38
7203495 c.1298T>G p.L433W Chr.8: 33500452-33500452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763002279 Chr.8: 33500411-33500411 on Build GRCh38
rs901561236 Chr.8: 33500223-33500223 on Build GRCh38
rs370553924 Chr.8: 33500416-33500416 on Build GRCh38
rs745366614 Chr.8: 33500365-33500365 on Build GRCh38
rs927679251 Chr.8: 33500311-33500311 on Build GRCh38
rs771891866 Chr.8: 33500378-33500378 on Build GRCh38
rs746607834 Chr.8: 33500385-33500385 on Build GRCh38
rs765248619 Chr.8: 33500284-33500284 on Build GRCh38
rs145382016 Chr.8: 33500355-33500355 on Build GRCh38
rs983789329 Chr.8: 33500219-33500219 on Build GRCh38
rs527301027 Chr.8: 33500245-33500245 on Build GRCh38
rs546375792 Chr.8: 33500296-33500296 on Build GRCh38
rs370105252 Chr.8: 33500398-33500398 on Build GRCh38
rs758541435 Chr.8: 33500399-33500400 on Build GRCh38
rs531387294 Chr.8: 33500350-33500350 on Build GRCh38
rs750505271 Chr.8: 33500291-33500291 on Build GRCh38
rs757207361 Chr.8: 33500434-33500434 on Build GRCh38
rs753651202 Chr.8: 33500432-33500432 on Build GRCh38
rs13439524 Chr.8: 33500300-33500300 on Build GRCh38
rs201577994 Chr.8: 33500351-33500351 on Build GRCh38
rs778344343 Chr.8: 33500360-33500360 on Build GRCh38
rs778878337 Chr.8: 33500438-33500438 on Build GRCh38
rs766398137 Chr.8: 33500415-33500415 on Build GRCh38
rs190298167 Chr.8: 33500336-33500336 on Build GRCh38
rs773323960 Chr.8: 33500399-33500399 on Build GRCh38
rs571647825 Chr.8: 33500393-33500393 on Build GRCh38
rs112755542 Chr.8: 33500359-33500359 on Build GRCh38
rs374210175 Chr.8: 33500418-33500418 on Build GRCh38
rs755926912 Chr.8: 33500335-33500335 on Build GRCh38
rs377112198 Chr.8: 33500356-33500356 on Build GRCh38
rs908208907 Chr.8: 33500270-33500270 on Build GRCh38
rs892791227 Chr.8: 33500384-33500384 on Build GRCh38
rs762339803 Chr.8: 33500282-33500282 on Build GRCh38
rs145192530 Chr.8: 33500394-33500394 on Build GRCh38
rs775257856 Chr.8: 33500382-33500382 on Build GRCh38

View Full Product Details