Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22884453-22884724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REM2
Entrez Gene ID
161253
Gene Name
RRAD and GEM like GTPase 2
Gene Aliases
-
UniGene
Hs.444911
Gene Chromosome Location
Chr.14: 22883165-22887680 on Build GRCh38
Transcripts
NM_173527.2 XM_005267383.3 XM_011536518.2 XM_017021058.1 XM_017021059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5597490 c.96G>A p.K32K Chr.14: 22884696-22884696 on Build GRCh38
3814503 c.96G>T p.K32N Chr.14: 22884696-22884696 on Build GRCh38
6764917 c.87G>A p.T29T Chr.14: 22884687-22884687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778534880 Chr.14: 22884679-22884679 on Build GRCh38
rs535015084 Chr.14: 22884603-22884603 on Build GRCh38
rs747385059 Chr.14: 22884688-22884688 on Build GRCh38
rs769877917 Chr.14: 22884493-22884493 on Build GRCh38
rs956168259 Chr.14: 22884503-22884503 on Build GRCh38
rs551589704 Chr.14: 22884607-22884607 on Build GRCh38
rs556296414 Chr.14: 22884631-22884631 on Build GRCh38
rs571675183 Chr.14: 22884619-22884619 on Build GRCh38
rs867235165 Chr.14: 22884671-22884671 on Build GRCh38
rs753885860 Chr.14: 22884638-22884638 on Build GRCh38
rs932775405 Chr.14: 22884494-22884494 on Build GRCh38
rs766956343 Chr.14: 22884629-22884629 on Build GRCh38
rs745313941 Chr.14: 22884684-22884684 on Build GRCh38
rs769063348 Chr.14: 22884696-22884696 on Build GRCh38
rs944530417 Chr.14: 22884482-22884482 on Build GRCh38
rs189717936 Chr.14: 22884511-22884511 on Build GRCh38
rs180792332 Chr.14: 22884620-22884620 on Build GRCh38
rs1040265632 Chr.14: 22884489-22884489 on Build GRCh38
rs201532289 Chr.14: 22884686-22884686 on Build GRCh38
rs757137970 Chr.14: 22884650-22884650 on Build GRCh38
rs964822420 Chr.14: 22884593-22884593 on Build GRCh38
rs45628834 Chr.14: 22884687-22884687 on Build GRCh38
rs920581822 Chr.14: 22884661-22884661 on Build GRCh38
rs549684908 Chr.14: 22884487-22884487 on Build GRCh38
rs776972071 Chr.14: 22884699-22884699 on Build GRCh38
rs529581069 Chr.14: 22884479-22884479 on Build GRCh38
rs911997529 Chr.14: 22884530-22884530 on Build GRCh38

View Full Product Details