Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37087207-37087456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC3
Entrez Gene ID
7267
Gene Name
tetratricopeptide repeat domain 3
Gene Aliases
DCRR1, RNF105, TPRDIII
UniGene
Hs.368214
Gene Chromosome Location
Chr.21: 37073184-37203118 on Build GRCh38
Transcripts
NM_001001894.1 NM_001320703.1 NM_001320704.1 NM_003316.3 XM_005261048.1 XM_005261049.1 XM_005261050.4 XM_005261052.1 XM_005261054.3 XM_005261055.1 XM_005261056.2 XM_011529740.2 XM_011529742.2 XM_017028458.1 XM_017028459.1 XM_017028460.1 XM_017028461.1 XM_017028462.1 XM_017028463.1 XM_017028466.1 XM_017028467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197977 c.7A>G p.N3D Chr.21: 37087264-37087264 on Build GRCh38
6446768 c.4G>T p.D2Y Chr.21: 37087261-37087261 on Build GRCh38
5697911 c.21A>T p.G7G Chr.21: 37087278-37087278 on Build GRCh38
3423946 c.107A>C p.D36A Chr.21: 37087364-37087364 on Build GRCh38
6313088 c.33G>T p.V11V Chr.21: 37087290-37087290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150569930 Chr.21: 37087323-37087323 on Build GRCh38
rs982960840 Chr.21: 37087341-37087341 on Build GRCh38
rs200452449 Chr.21: 37087288-37087288 on Build GRCh38
rs747537722 Chr.21: 37087260-37087260 on Build GRCh38
rs756896636 Chr.21: 37087407-37087407 on Build GRCh38
rs748588467 Chr.21: 37087335-37087335 on Build GRCh38
rs753775126 Chr.21: 37087264-37087264 on Build GRCh38
rs778461413 Chr.21: 37087410-37087410 on Build GRCh38
rs553316028 Chr.21: 37087267-37087267 on Build GRCh38
rs888486317 Chr.21: 37087384-37087384 on Build GRCh38
rs2835583 Chr.21: 37087425-37087425 on Build GRCh38
rs763813021 Chr.21: 37087397-37087397 on Build GRCh38
rs750009298 Chr.21: 37087417-37087417 on Build GRCh38
rs73201925 Chr.21: 37087293-37087293 on Build GRCh38
rs770057844 Chr.21: 37087337-37087337 on Build GRCh38
rs748900793 Chr.21: 37087322-37087322 on Build GRCh38
rs759449583 Chr.21: 37087369-37087369 on Build GRCh38
rs777552918 Chr.21: 37087320-37087320 on Build GRCh38
rs199612355 Chr.21: 37087373-37087373 on Build GRCh38
rs908752273 Chr.21: 37087358-37087358 on Build GRCh38
rs375612625 Chr.21: 37087244-37087244 on Build GRCh38
rs141884251 Chr.21: 37087319-37087319 on Build GRCh38
rs866363802 Chr.21: 37087301-37087301 on Build GRCh38
rs565611168 Chr.21: 37087290-37087292 on Build GRCh38
rs930252432 Chr.21: 37087332-37087332 on Build GRCh38
rs374057147 Chr.21: 37087426-37087426 on Build GRCh38
rs767269766 Chr.21: 37087306-37087306 on Build GRCh38
rs767504600 Chr.21: 37087372-37087372 on Build GRCh38
rs773160877 Chr.21: 37087277-37087277 on Build GRCh38
rs563231547 Chr.21: 37087424-37087424 on Build GRCh38
rs540038399 Chr.21: 37087265-37087265 on Build GRCh38
rs760400196 Chr.21: 37087388-37087388 on Build GRCh38
rs754941611 Chr.21: 37087299-37087299 on Build GRCh38
rs772456933 Chr.21: 37087247-37087247 on Build GRCh38
rs200604513 Chr.21: 37087385-37087385 on Build GRCh38
rs201528757 Chr.21: 37087346-37087346 on Build GRCh38
rs144396837 Chr.21: 37087401-37087401 on Build GRCh38
rs763107912 Chr.21: 37087344-37087344 on Build GRCh38
rs769099564 Chr.21: 37087261-37087261 on Build GRCh38
rs199602707 Chr.21: 37087416-37087416 on Build GRCh38
rs148592918 Chr.21: 37087329-37087329 on Build GRCh38
rs766442874 Chr.21: 37087351-37087351 on Build GRCh38
rs769332677 Chr.21: 37087328-37087328 on Build GRCh38
rs776001773 Chr.21: 37087258-37087258 on Build GRCh38
rs774456054 Chr.21: 37087361-37087361 on Build GRCh38
rs377754447 Chr.21: 37087318-37087318 on Build GRCh38
rs144830241 Chr.21: 37087324-37087324 on Build GRCh38
rs757099449 Chr.21: 37087289-37087290 on Build GRCh38
rs766382758 Chr.21: 37087290-37087290 on Build GRCh38
rs765302589 Chr.21: 37087402-37087402 on Build GRCh38
rs773425574 Chr.21: 37087339-37087339 on Build GRCh38

View Full Product Details