Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7694823-7695096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OVCH2
Entrez Gene ID
341277
Gene Name
ovochymase 2 (gene/pseudogene)
Gene Aliases
OVTN
UniGene
Hs.532475
Gene Chromosome Location
Chr.11: 7678623-7706894 on Build GRCh38
Transcripts
NM_198185.4 XM_017017659.1

SNP Detail

SNP ID Chromosome Location
rs187563006 Chr.11: 7694874-7694874 on Build GRCh38
rs968208655 Chr.11: 7695006-7695006 on Build GRCh38
rs906911795 Chr.11: 7694935-7694935 on Build GRCh38
rs986270993 Chr.11: 7695014-7695014 on Build GRCh38
rs1022509322 Chr.11: 7694992-7694992 on Build GRCh38
rs892737142 Chr.11: 7694950-7694950 on Build GRCh38
rs267603203 Chr.11: 7695069-7695069 on Build GRCh38
rs1040354129 Chr.11: 7694869-7694869 on Build GRCh38
rs770300536 Chr.11: 7695025-7695025 on Build GRCh38
rs547015579 Chr.11: 7694914-7694914 on Build GRCh38
rs368286255 Chr.11: 7694897-7694897 on Build GRCh38
rs910608206 Chr.11: 7694860-7694860 on Build GRCh38
rs986893191 Chr.11: 7694871-7694871 on Build GRCh38
rs11602675 Chr.11: 7694997-7694997 on Build GRCh38
rs944719237 Chr.11: 7694868-7694868 on Build GRCh38
rs919257762 Chr.11: 7695042-7695042 on Build GRCh38
rs571319662 Chr.11: 7694926-7694926 on Build GRCh38
rs750365489 Chr.11: 7694947-7694947 on Build GRCh38
rs895288556 Chr.11: 7695022-7695022 on Build GRCh38
rs1054351369 Chr.11: 7694888-7694888 on Build GRCh38
rs193298737 Chr.11: 7694915-7694915 on Build GRCh38
rs929897818 Chr.11: 7694876-7694876 on Build GRCh38
rs1017124316 Chr.11: 7695030-7695030 on Build GRCh38
rs564222518 Chr.11: 7694949-7694949 on Build GRCh38
rs200281458 Chr.11: 7694903-7694903 on Build GRCh38
rs544267992 Chr.11: 7694923-7694923 on Build GRCh38
rs766778850 Chr.11: 7695036-7695036 on Build GRCh38
rs773524170 Chr.11: 7695034-7695034 on Build GRCh38
rs1055486046 Chr.11: 7695019-7695019 on Build GRCh38
rs904911419 Chr.11: 7694919-7694919 on Build GRCh38
rs146967963 Chr.11: 7694883-7694883 on Build GRCh38
rs941158962 Chr.11: 7694907-7694907 on Build GRCh38
rs774991933 Chr.11: 7695056-7695056 on Build GRCh38
rs1009812300 Chr.11: 7694965-7694965 on Build GRCh38
rs575272752 Chr.11: 7694855-7694856 on Build GRCh38
rs527634539 Chr.11: 7694938-7694938 on Build GRCh38
rs80197536 Chr.11: 7695035-7695035 on Build GRCh38
rs911020692 Chr.11: 7694877-7694877 on Build GRCh38

View Full Product Details