Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62361491-62361764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF11A
Entrez Gene ID
8792
Gene Name
TNF receptor superfamily member 11a
Gene Aliases
CD265, FEO, LOH18CR1, ODFR, OFE, OPTB7, OSTS, PDB2, RANK, TRANCER
UniGene
Hs.204044
Gene Chromosome Location
Chr.18: 62325287-62388096 on Build GRCh38
Transcripts
NM_001270949.1 NM_001270950.1 NM_001270951.1 NM_001278268.1 NM_003839.3 XM_011526244.2 XM_011526245.2 XM_017026064.1 XM_017026065.1 XM_017026066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686986 c.618A>G p.E206E Chr.18: 62361681-62361681 on Build GRCh38
6320637 c.678G>T p.V226V Chr.18: 62361741-62361741 on Build GRCh38
1288533 c.662C>T p.A221V Chr.18: 62361725-62361725 on Build GRCh38
709171 c.661G>A p.A221T Chr.18: 62361724-62361724 on Build GRCh38
3937776 c.697G>A p.V233I Chr.18: 62361760-62361760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537966416 Chr.18: 62361724-62361724 on Build GRCh38
rs749743154 Chr.18: 62361733-62361733 on Build GRCh38
rs562681425 Chr.18: 62361523-62361523 on Build GRCh38
rs561759882 Chr.18: 62361608-62361608 on Build GRCh38
rs766330561 Chr.18: 62361700-62361700 on Build GRCh38
rs753664199 Chr.18: 62361716-62361716 on Build GRCh38
rs35407865 Chr.18: 62361588-62361588 on Build GRCh38
rs372839394 Chr.18: 62361654-62361654 on Build GRCh38
rs975853382 Chr.18: 62361611-62361611 on Build GRCh38
rs568097303 Chr.18: 62361731-62361731 on Build GRCh38
rs17069895 Chr.18: 62361625-62361625 on Build GRCh38
rs34591321 Chr.18: 62361660-62361660 on Build GRCh38
rs76477458 Chr.18: 62361609-62361609 on Build GRCh38
rs562883523 Chr.18: 62361573-62361573 on Build GRCh38
rs994798457 Chr.18: 62361527-62361527 on Build GRCh38
rs1009039225 Chr.18: 62361557-62361557 on Build GRCh38
rs761515551 Chr.18: 62361675-62361675 on Build GRCh38
rs71352572 Chr.18: 62361518-62361518 on Build GRCh38
rs138444094 Chr.18: 62361726-62361726 on Build GRCh38
rs541813708 Chr.18: 62361596-62361596 on Build GRCh38
rs978381515 Chr.18: 62361634-62361634 on Build GRCh38
rs1020040832 Chr.18: 62361548-62361548 on Build GRCh38
rs548522707 Chr.18: 62361550-62361551 on Build GRCh38
rs769838090 Chr.18: 62361662-62361662 on Build GRCh38
rs907952727 Chr.18: 62361628-62361628 on Build GRCh38
rs781286218 Chr.18: 62361648-62361648 on Build GRCh38
rs1013649856 Chr.18: 62361514-62361514 on Build GRCh38
rs952950572 Chr.18: 62361547-62361547 on Build GRCh38
rs770301582 Chr.18: 62361657-62361657 on Build GRCh38
rs756637700 Chr.18: 62361727-62361727 on Build GRCh38
rs781169173 Chr.18: 62361725-62361725 on Build GRCh38
rs769081017 Chr.18: 62361669-62361669 on Build GRCh38
rs769177863 Chr.18: 62361539-62361539 on Build GRCh38
rs1019042504 Chr.18: 62361566-62361566 on Build GRCh38
rs774147578 Chr.18: 62361670-62361670 on Build GRCh38
rs772749823 Chr.18: 62361691-62361691 on Build GRCh38
rs146793660 Chr.18: 62361688-62361688 on Build GRCh38
rs922908221 Chr.18: 62361616-62361616 on Build GRCh38
rs746265199 Chr.18: 62361668-62361668 on Build GRCh38
rs200341009 Chr.18: 62361721-62361721 on Build GRCh38
rs534076974 Chr.18: 62361723-62361723 on Build GRCh38
rs757358860 Chr.18: 62361635-62361635 on Build GRCh38
rs376762500 Chr.18: 62361526-62361526 on Build GRCh38
rs375075146 Chr.18: 62361699-62361699 on Build GRCh38
rs773208145 Chr.18: 62361542-62361542 on Build GRCh38
rs8099222 Chr.18: 62361529-62361529 on Build GRCh38

View Full Product Details