Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43962640-43962905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF221
Entrez Gene ID
7638
Gene Name
zinc finger protein 221
Gene Aliases
-
UniGene
Hs.631598
Gene Chromosome Location
Chr.19: 43951196-43970207 on Build GRCh38
Transcripts
NM_001297588.1 NM_001297589.1 NM_013359.2 XM_017027232.1 XM_017027233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409578 c.17T>G p.L6R Chr.19: 43962743-43962743 on Build GRCh38
3535379 c.49C>T p.P17S Chr.19: 43962775-43962775 on Build GRCh38
6317635 c.19G>T p.E7* Chr.19: 43962745-43962745 on Build GRCh38
1394348 c.37_38delCT p.C14fs*4 Chr.19: 43962763-43962764 on Build GRCh38
1612369 c.3G>A p.M1I Chr.19: 43962729-43962729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020804650 Chr.19: 43962865-43962865 on Build GRCh38
rs897165415 Chr.19: 43962692-43962692 on Build GRCh38
rs866069705 Chr.19: 43962812-43962812 on Build GRCh38
rs368170959 Chr.19: 43962748-43962748 on Build GRCh38
rs779243890 Chr.19: 43962798-43962798 on Build GRCh38
rs368188890 Chr.19: 43962815-43962815 on Build GRCh38
rs755185106 Chr.19: 43962714-43962714 on Build GRCh38
rs373675846 Chr.19: 43962725-43962725 on Build GRCh38
rs747754239 Chr.19: 43962704-43962704 on Build GRCh38
rs562637173 Chr.19: 43962734-43962734 on Build GRCh38
rs780865940 Chr.19: 43962691-43962691 on Build GRCh38
rs150669301 Chr.19: 43962766-43962766 on Build GRCh38
rs780956282 Chr.19: 43962763-43962764 on Build GRCh38
rs376412812 Chr.19: 43962719-43962719 on Build GRCh38
rs759995605 Chr.19: 43962834-43962834 on Build GRCh38
rs113081238 Chr.19: 43962858-43962858 on Build GRCh38
rs150413608 Chr.19: 43962669-43962669 on Build GRCh38
rs138913252 Chr.19: 43962802-43962802 on Build GRCh38
rs956449407 Chr.19: 43962853-43962853 on Build GRCh38
rs750167618 Chr.19: 43962773-43962773 on Build GRCh38
rs370296511 Chr.19: 43962720-43962720 on Build GRCh38
rs769517591 Chr.19: 43962705-43962705 on Build GRCh38
rs779719793 Chr.19: 43962789-43962789 on Build GRCh38
rs758282806 Chr.19: 43962778-43962778 on Build GRCh38
rs146575991 Chr.19: 43962800-43962800 on Build GRCh38
rs767535369 Chr.19: 43962740-43962740 on Build GRCh38
rs370636079 Chr.19: 43962761-43962761 on Build GRCh38
rs575228648 Chr.19: 43962780-43962780 on Build GRCh38
rs533062889 Chr.19: 43962833-43962833 on Build GRCh38
rs185269639 Chr.19: 43962831-43962831 on Build GRCh38
rs769423971 Chr.19: 43962834-43962834 on Build GRCh38
rs540789766 Chr.19: 43962881-43962881 on Build GRCh38
rs748896578 Chr.19: 43962823-43962823 on Build GRCh38
rs771268052 Chr.19: 43962829-43962829 on Build GRCh38
rs1039888375 Chr.19: 43962776-43962776 on Build GRCh38
rs374341238 Chr.19: 43962785-43962785 on Build GRCh38
rs745704720 Chr.19: 43962800-43962803 on Build GRCh38
rs1007542884 Chr.19: 43962764-43962764 on Build GRCh38
rs949686739 Chr.19: 43962668-43962668 on Build GRCh38

View Full Product Details