Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33286914-33287185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR46
Entrez Gene ID
9277
Gene Name
WD repeat domain 46
Gene Aliases
BING4, C6orf11, FP221, UTP7
UniGene
Hs.520063
Gene Chromosome Location
Chr.6: 33278850-33289527 on Build GRCh38
Transcripts
NM_001164267.1 NM_005452.5 XM_017011484.1 XM_017011485.1 XM_017011486.1 XM_017011487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4789632 c.998A>G p.H333R Chr.6: 33287108-33287108 on Build GRCh38
7146323 c.946C>A p.R316R Chr.6: 33287160-33287160 on Build GRCh38
3724172 c.990C>T p.A330A Chr.6: 33287116-33287116 on Build GRCh38
7079180 c.939G>A p.L313L Chr.6: 33287167-33287167 on Build GRCh38
291650 c.961G>A p.D321N Chr.6: 33287145-33287145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145596280 Chr.6: 33287150-33287150 on Build GRCh38
rs756346756 Chr.6: 33287091-33287091 on Build GRCh38
rs997207761 Chr.6: 33287094-33287094 on Build GRCh38
rs755355646 Chr.6: 33287115-33287115 on Build GRCh38
rs754818056 Chr.6: 33287064-33287065 on Build GRCh38
rs746685125 Chr.6: 33287122-33287122 on Build GRCh38
rs761809733 Chr.6: 33286943-33286943 on Build GRCh38
rs547526632 Chr.6: 33287029-33287029 on Build GRCh38
rs372460246 Chr.6: 33287069-33287069 on Build GRCh38
rs529857192 Chr.6: 33287098-33287098 on Build GRCh38
rs1030441297 Chr.6: 33287100-33287100 on Build GRCh38
rs772814628 Chr.6: 33286957-33286957 on Build GRCh38
rs775292869 Chr.6: 33287145-33287145 on Build GRCh38
rs765486438 Chr.6: 33286954-33286954 on Build GRCh38
rs748633871 Chr.6: 33287117-33287117 on Build GRCh38
rs777335544 Chr.6: 33287119-33287119 on Build GRCh38
rs201100313 Chr.6: 33287043-33287043 on Build GRCh38
rs117463878 Chr.6: 33287116-33287116 on Build GRCh38
rs901607202 Chr.6: 33287022-33287022 on Build GRCh38
rs192701143 Chr.6: 33287142-33287142 on Build GRCh38
rs367822545 Chr.6: 33287026-33287026 on Build GRCh38
rs559194115 Chr.6: 33287038-33287038 on Build GRCh38
rs1007231609 Chr.6: 33286982-33286982 on Build GRCh38
rs747309051 Chr.6: 33287032-33287032 on Build GRCh38
rs374221252 Chr.6: 33287024-33287024 on Build GRCh38
rs1016069066 Chr.6: 33287139-33287139 on Build GRCh38
rs1016183002 Chr.6: 33287135-33287135 on Build GRCh38
rs780771980 Chr.6: 33287019-33287019 on Build GRCh38
rs142670301 Chr.6: 33287104-33287104 on Build GRCh38
rs375694480 Chr.6: 33287140-33287140 on Build GRCh38
rs770535669 Chr.6: 33287124-33287124 on Build GRCh38
rs371734662 Chr.6: 33287160-33287160 on Build GRCh38
rs773319470 Chr.6: 33287146-33287146 on Build GRCh38
rs1026189003 Chr.6: 33287103-33287103 on Build GRCh38
rs757854724 Chr.6: 33287118-33287118 on Build GRCh38
rs779094358 Chr.6: 33287086-33287087 on Build GRCh38
rs776222936 Chr.6: 33287127-33287127 on Build GRCh38
rs918162346 Chr.6: 33287021-33287021 on Build GRCh38

View Full Product Details