Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151185673-151185946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS72
Entrez Gene ID
6944
Gene Name
vacuolar protein sorting 72 homolog
Gene Aliases
CFL1, Swc2, TCFL1, YL-1, YL1
UniGene
Hs.2430
Gene Chromosome Location
Chr.1: 151176300-151190213 on Build GRCh38
Transcripts
NM_001271087.1 NM_001271088.1 NM_005997.2 XM_017002205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3934087 c.248G>A p.R83Q Chr.1: 151185820-151185820 on Build GRCh38
1333946 c.244C>T p.R82C Chr.1: 151185824-151185824 on Build GRCh38
7095230 c.133G>A p.E45K Chr.1: 151185935-151185935 on Build GRCh38
458603 c.218G>C p.G73A Chr.1: 151185850-151185850 on Build GRCh38
1333947 c.150G>A p.Q50Q Chr.1: 151185918-151185918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776748360 Chr.1: 151185824-151185824 on Build GRCh38
rs756727605 Chr.1: 151185903-151185903 on Build GRCh38
rs753341635 Chr.1: 151185787-151185787 on Build GRCh38
rs775467288 Chr.1: 151185869-151185869 on Build GRCh38
rs746165229 Chr.1: 151185792-151185792 on Build GRCh38
rs965702209 Chr.1: 151185823-151185823 on Build GRCh38
rs367758804 Chr.1: 151185820-151185820 on Build GRCh38
rs767405152 Chr.1: 151185858-151185858 on Build GRCh38
rs1048664320 Chr.1: 151185743-151185743 on Build GRCh38
rs144454139 Chr.1: 151185882-151185882 on Build GRCh38
rs760640116 Chr.1: 151185876-151185876 on Build GRCh38
rs773571919 Chr.1: 151185837-151185837 on Build GRCh38
rs953479161 Chr.1: 151185737-151185737 on Build GRCh38
rs753348679 Chr.1: 151185885-151185885 on Build GRCh38
rs759345946 Chr.1: 151185846-151185846 on Build GRCh38
rs781737774 Chr.1: 151185776-151185778 on Build GRCh38
rs757325748 Chr.1: 151185802-151185802 on Build GRCh38
rs769082492 Chr.1: 151185821-151185821 on Build GRCh38
rs775748824 Chr.1: 151185795-151185795 on Build GRCh38
rs770017421 Chr.1: 151185835-151185835 on Build GRCh38
rs778465301 Chr.1: 151185791-151185791 on Build GRCh38
rs759431818 Chr.1: 151185755-151185755 on Build GRCh38
rs373691223 Chr.1: 151185752-151185752 on Build GRCh38
rs200216847 Chr.1: 151185851-151185851 on Build GRCh38
rs754334268 Chr.1: 151185734-151185734 on Build GRCh38
rs587742534 Chr.1: 151185788-151185788 on Build GRCh38
rs764815849 Chr.1: 151185913-151185913 on Build GRCh38
rs753649179 Chr.1: 151185746-151185746 on Build GRCh38
rs780456410 Chr.1: 151185816-151185816 on Build GRCh38
rs767492745 Chr.1: 151185770-151185770 on Build GRCh38
rs752691500 Chr.1: 151185771-151185771 on Build GRCh38
rs756176679 Chr.1: 151185777-151185777 on Build GRCh38
rs777308768 Chr.1: 151185781-151185781 on Build GRCh38

View Full Product Details