Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231795867-231796140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7B
Entrez Gene ID
64708
Gene Name
COP9 signalosome subunit 7B
Gene Aliases
CSN7B, SGN7b
UniGene
Hs.335061
Gene Chromosome Location
Chr.2: 231781671-231809253 on Build GRCh38
Transcripts
NM_001282949.2 NM_001282950.2 NM_001282951.2 NM_001282952.2 NM_001308381.1 NM_022730.3 XM_006712693.1 XM_011511638.1 XM_011511639.1 XM_011511640.1 XM_017004716.1 XM_017004717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391564 c.342C>T p.S114S Chr.2: 231796120-231796120 on Build GRCh38
3713636 c.351G>C p.L117L Chr.2: 231796129-231796129 on Build GRCh38
5396470 c.341C>T p.S114F Chr.2: 231796119-231796119 on Build GRCh38
3579042 c.358C>T p.L120L Chr.2: 231796136-231796136 on Build GRCh38
4517397 c.334_335CC>TT p.P112F Chr.2: 231796112-231796113 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749652282 Chr.2: 231796099-231796099 on Build GRCh38
rs900643077 Chr.2: 231795914-231795914 on Build GRCh38
rs934891103 Chr.2: 231795931-231795931 on Build GRCh38
rs537826694 Chr.2: 231795992-231795992 on Build GRCh38
rs552275128 Chr.2: 231795901-231795901 on Build GRCh38
rs993129598 Chr.2: 231796090-231796090 on Build GRCh38
rs536505292 Chr.2: 231796002-231796002 on Build GRCh38
rs765850808 Chr.2: 231796051-231796056 on Build GRCh38
rs555258827 Chr.2: 231796008-231796008 on Build GRCh38
rs73088818 Chr.2: 231796070-231796070 on Build GRCh38
rs200036781 Chr.2: 231796111-231796111 on Build GRCh38
rs367651903 Chr.2: 231796057-231796057 on Build GRCh38
rs537451982 Chr.2: 231796032-231796032 on Build GRCh38
rs570591098 Chr.2: 231795975-231795975 on Build GRCh38
rs754700543 Chr.2: 231796117-231796117 on Build GRCh38
rs754289423 Chr.2: 231796072-231796072 on Build GRCh38
rs879151697 Chr.2: 231795991-231795991 on Build GRCh38
rs144978316 Chr.2: 231796120-231796120 on Build GRCh38
rs917450723 Chr.2: 231796093-231796093 on Build GRCh38
rs573072648 Chr.2: 231796019-231796019 on Build GRCh38
rs770251188 Chr.2: 231795940-231795940 on Build GRCh38
rs1057500304 Chr.2: 231795956-231795956 on Build GRCh38
rs758677034 Chr.2: 231796090-231796091 on Build GRCh38
rs757699495 Chr.2: 231796029-231796029 on Build GRCh38
rs751046114 Chr.2: 231796103-231796103 on Build GRCh38
rs777438775 Chr.2: 231796087-231796087 on Build GRCh38
rs756820867 Chr.2: 231796107-231796107 on Build GRCh38
rs761901610 Chr.2: 231796058-231796058 on Build GRCh38
rs184987484 Chr.2: 231795994-231795994 on Build GRCh38
rs773223109 Chr.2: 231796061-231796061 on Build GRCh38
rs376612825 Chr.2: 231796081-231796081 on Build GRCh38
rs745763588 Chr.2: 231796114-231796114 on Build GRCh38
rs757934865 Chr.2: 231796085-231796085 on Build GRCh38
rs769454878 Chr.2: 231796056-231796056 on Build GRCh38
rs761179083 Chr.2: 231796063-231796063 on Build GRCh38
rs772162511 Chr.2: 231796059-231796059 on Build GRCh38
rs59929623 Chr.2: 231796055-231796055 on Build GRCh38
rs558610154 Chr.2: 231796054-231796054 on Build GRCh38
rs759991756 Chr.2: 231796073-231796073 on Build GRCh38
rs368066879 Chr.2: 231796082-231796082 on Build GRCh38
rs746213114 Chr.2: 231796079-231796079 on Build GRCh38
rs1019268643 Chr.2: 231796049-231796049 on Build GRCh38
rs571637225 Chr.2: 231796001-231796001 on Build GRCh38

View Full Product Details