Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25675202-25675463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF337
Entrez Gene ID
26152
Gene Name
zinc finger protein 337
Gene Aliases
-
UniGene
Hs.61881
Gene Chromosome Location
Chr.20: 25673195-25696904 on Build GRCh38
Transcripts
NM_001290261.1 NM_015655.3 XM_005260702.3 XM_006723558.3 XM_011529219.2 XM_017027802.1 XM_017027803.1 XM_017027804.1 NR_126465.1

Gene Symbol
ZNF337-AS1
Entrez Gene ID
102724826
Gene Name
ZNF337 antisense RNA 1
Gene Aliases
-
UniGene
Hs.633621
Gene Chromosome Location
Chr.20: 25624045-25678074 on Build GRCh38
Transcripts
NM_001290261.1 NM_015655.3 XM_005260702.3 XM_006723558.3 XM_011529219.2 XM_017027802.1 XM_017027803.1 XM_017027804.1 NR_126465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4097309 c.2028G>T p.W676C Chr.20: 25675260-25675260 on Build GRCh38
1751363 c.1936C>T p.H646Y Chr.20: 25675352-25675352 on Build GRCh38
2887666 c.1903C>T p.R635* Chr.20: 25675385-25675385 on Build GRCh38
4942086 c.2037C>G p.H679Q Chr.20: 25675251-25675251 on Build GRCh38
165802 c.1904G>A p.R635Q Chr.20: 25675384-25675384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766505797 Chr.20: 25675424-25675424 on Build GRCh38
rs935041321 Chr.20: 25675269-25675269 on Build GRCh38
rs754710429 Chr.20: 25675431-25675431 on Build GRCh38
rs953422638 Chr.20: 25675420-25675420 on Build GRCh38
rs761961207 Chr.20: 25675279-25675279 on Build GRCh38
rs1029039919 Chr.20: 25675396-25675396 on Build GRCh38
rs770472681 Chr.20: 25675360-25675360 on Build GRCh38
rs376902779 Chr.20: 25675326-25675326 on Build GRCh38
rs140472566 Chr.20: 25675258-25675258 on Build GRCh38
rs747708143 Chr.20: 25675334-25675334 on Build GRCh38
rs769253992 Chr.20: 25675384-25675384 on Build GRCh38
rs755177493 Chr.20: 25675356-25675358 on Build GRCh38
rs751728909 Chr.20: 25675265-25675265 on Build GRCh38
rs771755693 Chr.20: 25675228-25675228 on Build GRCh38
rs150351850 Chr.20: 25675300-25675300 on Build GRCh38
rs1009380190 Chr.20: 25675251-25675251 on Build GRCh38
rs775871346 Chr.20: 25675259-25675259 on Build GRCh38
rs756131441 Chr.20: 25675334-25675335 on Build GRCh38
rs780006029 Chr.20: 25675338-25675339 on Build GRCh38
rs866179926 Chr.20: 25675357-25675357 on Build GRCh38
rs1030865368 Chr.20: 25675380-25675380 on Build GRCh38
rs770630580 Chr.20: 25675308-25675308 on Build GRCh38
rs146126391 Chr.20: 25675319-25675319 on Build GRCh38
rs760179219 Chr.20: 25675271-25675271 on Build GRCh38
rs765184760 Chr.20: 25675433-25675433 on Build GRCh38
rs772831889 Chr.20: 25675237-25675237 on Build GRCh38
rs140114787 Chr.20: 25675320-25675320 on Build GRCh38
rs1043749028 Chr.20: 25675339-25675339 on Build GRCh38
rs1043876055 Chr.20: 25675225-25675225 on Build GRCh38
rs763195980 Chr.20: 25675381-25675381 on Build GRCh38
rs746612198 Chr.20: 25675358-25675358 on Build GRCh38
rs147908423 Chr.20: 25675238-25675238 on Build GRCh38
rs761016141 Chr.20: 25675397-25675397 on Build GRCh38
rs777403937 Chr.20: 25675347-25675347 on Build GRCh38
rs764500458 Chr.20: 25675263-25675263 on Build GRCh38
rs773583861 Chr.20: 25675391-25675391 on Build GRCh38
rs925463115 Chr.20: 25675233-25675233 on Build GRCh38
rs767873385 Chr.20: 25675280-25675280 on Build GRCh38
rs200053406 Chr.20: 25675388-25675388 on Build GRCh38
rs771524582 Chr.20: 25675344-25675344 on Build GRCh38
rs753712062 Chr.20: 25675430-25675430 on Build GRCh38
rs767640187 Chr.20: 25675390-25675390 on Build GRCh38
rs750492365 Chr.20: 25675331-25675331 on Build GRCh38
rs999821969 Chr.20: 25675345-25675345 on Build GRCh38
rs202001008 Chr.20: 25675385-25675385 on Build GRCh38
rs754051803 Chr.20: 25675350-25675351 on Build GRCh38
rs6076383 Chr.20: 25675298-25675298 on Build GRCh38
rs903788651 Chr.20: 25675340-25675340 on Build GRCh38
rs775985363 Chr.20: 25675379-25675379 on Build GRCh38
rs200228007 Chr.20: 25675297-25675297 on Build GRCh38
rs373233055 Chr.20: 25675392-25675392 on Build GRCh38
rs752929963 Chr.20: 25675327-25675327 on Build GRCh38

View Full Product Details