Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3656974-3657245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1
Entrez Gene ID
1773
Gene Name
deoxyribonuclease I
Gene Aliases
DNL1, DRNI
UniGene
Hs.629638
Gene Chromosome Location
Chr.16: 3611705-3665472 on Build GRCh38
Transcripts
NM_005223.3 XM_005255148.3 XM_006720854.3 XM_011522393.2 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022996.1 XM_017022997.1 XM_017022998.1 XM_017022999.1 XM_017023000.1 XM_017023001.1 XM_017023002.1 XM_017023003.1 XM_017023004.1 XM_017023005.1 XM_017023006.1 XM_017023007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473744 c.565G>T p.G189C Chr.16: 3657202-3657202 on Build GRCh38
3746415 c.593T>G p.V198G Chr.16: 3657230-3657230 on Build GRCh38
1377795 c.513C>T p.D171D Chr.16: 3657075-3657075 on Build GRCh38
5775141 c.550-8C>G p.? Chr.16: 3657179-3657179 on Build GRCh38
5980905 c.499G>C p.D167H Chr.16: 3657061-3657061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1799891 Chr.16: 3657022-3657022 on Build GRCh38
rs8176925 Chr.16: 3657163-3657163 on Build GRCh38
rs535355164 Chr.16: 3657055-3657055 on Build GRCh38
rs777609844 Chr.16: 3657017-3657017 on Build GRCh38
rs773757254 Chr.16: 3657130-3657130 on Build GRCh38
rs770253517 Chr.16: 3657128-3657128 on Build GRCh38
rs969348584 Chr.16: 3657084-3657084 on Build GRCh38
rs191253160 Chr.16: 3657127-3657127 on Build GRCh38
rs140745748 Chr.16: 3657076-3657076 on Build GRCh38
rs201205627 Chr.16: 3657045-3657045 on Build GRCh38
rs746208858 Chr.16: 3657218-3657218 on Build GRCh38
rs371345873 Chr.16: 3657064-3657064 on Build GRCh38
rs539275502 Chr.16: 3657144-3657144 on Build GRCh38
rs747197899 Chr.16: 3657117-3657117 on Build GRCh38
rs552883126 Chr.16: 3657041-3657041 on Build GRCh38
rs368307903 Chr.16: 3657007-3657007 on Build GRCh38
rs751150786 Chr.16: 3657046-3657046 on Build GRCh38
rs563863449 Chr.16: 3657006-3657006 on Build GRCh38
rs908137861 Chr.16: 3657102-3657102 on Build GRCh38
rs777126816 Chr.16: 3657067-3657067 on Build GRCh38
rs143371936 Chr.16: 3657215-3657215 on Build GRCh38
rs763381140 Chr.16: 3657037-3657038 on Build GRCh38
rs748663246 Chr.16: 3657072-3657072 on Build GRCh38
rs757292367 Chr.16: 3657105-3657105 on Build GRCh38
rs369279602 Chr.16: 3657132-3657132 on Build GRCh38
rs139424576 Chr.16: 3657061-3657061 on Build GRCh38
rs765308996 Chr.16: 3657108-3657108 on Build GRCh38
rs973626714 Chr.16: 3657114-3657114 on Build GRCh38
rs751278286 Chr.16: 3657195-3657195 on Build GRCh38
rs755627104 Chr.16: 3657054-3657054 on Build GRCh38
rs377081589 Chr.16: 3657164-3657164 on Build GRCh38
rs1004576655 Chr.16: 3657179-3657179 on Build GRCh38
rs112247144 Chr.16: 3657087-3657087 on Build GRCh38
rs143407371 Chr.16: 3657032-3657032 on Build GRCh38
rs377278126 Chr.16: 3657039-3657039 on Build GRCh38
rs758328706 Chr.16: 3657112-3657112 on Build GRCh38
rs752220090 Chr.16: 3657053-3657053 on Build GRCh38
rs776488257 Chr.16: 3657186-3657186 on Build GRCh38
rs917296955 Chr.16: 3657124-3657124 on Build GRCh38
rs147093089 Chr.16: 3657193-3657193 on Build GRCh38
rs756241944 Chr.16: 3657171-3657171 on Build GRCh38
rs375288137 Chr.16: 3657175-3657175 on Build GRCh38
rs763527559 Chr.16: 3657091-3657091 on Build GRCh38
rs566411600 Chr.16: 3657044-3657044 on Build GRCh38
rs769436738 Chr.16: 3657189-3657189 on Build GRCh38
rs781150236 Chr.16: 3657125-3657125 on Build GRCh38
rs780928961 Chr.16: 3657225-3657225 on Build GRCh38
rs753781014 Chr.16: 3657101-3657101 on Build GRCh38
rs775126039 Chr.16: 3657150-3657150 on Build GRCh38
rs916693567 Chr.16: 3657011-3657011 on Build GRCh38
rs772557385 Chr.16: 3657221-3657221 on Build GRCh38
rs771265621 Chr.16: 3657143-3657143 on Build GRCh38
rs770091885 Chr.16: 3657075-3657075 on Build GRCh38
rs746102135 Chr.16: 3657030-3657030 on Build GRCh38
rs372063579 Chr.16: 3657024-3657024 on Build GRCh38
rs758143421 Chr.16: 3657158-3657158 on Build GRCh38
rs773927363 Chr.16: 3657009-3657010 on Build GRCh38
rs752528209 Chr.16: 3657003-3657003 on Build GRCh38
rs768175110 Chr.16: 3657185-3657185 on Build GRCh38
rs766818831 Chr.16: 3657058-3657059 on Build GRCh38
rs8176932 Chr.16: 3657159-3657159 on Build GRCh38
rs779212028 Chr.16: 3657029-3657029 on Build GRCh38
rs146238243 Chr.16: 3657038-3657038 on Build GRCh38
rs780321271 Chr.16: 3657113-3657113 on Build GRCh38
rs755025011 Chr.16: 3657122-3657122 on Build GRCh38
rs146236198 Chr.16: 3657212-3657212 on Build GRCh38
rs8176938 Chr.16: 3657090-3657090 on Build GRCh38
rs370513422 Chr.16: 3657174-3657174 on Build GRCh38
rs143161590 Chr.16: 3656991-3656991 on Build GRCh38
rs746285287 Chr.16: 3657184-3657184 on Build GRCh38
rs59621760 Chr.16: 3657063-3657063 on Build GRCh38
rs760269041 Chr.16: 3657147-3657147 on Build GRCh38
rs375072190 Chr.16: 3657139-3657139 on Build GRCh38
rs142644209 Chr.16: 3657205-3657205 on Build GRCh38
rs920573024 Chr.16: 3657213-3657213 on Build GRCh38
rs201942334 Chr.16: 3657060-3657060 on Build GRCh38
rs369538834 Chr.16: 3657151-3657151 on Build GRCh38
rs780689531 Chr.16: 3656997-3656997 on Build GRCh38
rs74892550 Chr.16: 3657190-3657190 on Build GRCh38
rs759204609 Chr.16: 3657049-3657049 on Build GRCh38
rs751121067 Chr.16: 3657221-3657223 on Build GRCh38
rs751277123 Chr.16: 3656992-3656992 on Build GRCh38
rs1026535839 Chr.16: 3657047-3657047 on Build GRCh38
rs781516300 Chr.16: 3657165-3657165 on Build GRCh38
rs34984360 Chr.16: 3657216-3657216 on Build GRCh38
rs754414801 Chr.16: 3656996-3656996 on Build GRCh38
rs8175352 Chr.16: 3657204-3657204 on Build GRCh38
rs761406088 Chr.16: 3657188-3657188 on Build GRCh38
rs775674591 Chr.16: 3657033-3657033 on Build GRCh38
rs376993853 Chr.16: 3657153-3657153 on Build GRCh38
rs539405862 Chr.16: 3657183-3657183 on Build GRCh38
rs775061418 Chr.16: 3657088-3657088 on Build GRCh38
rs199540176 Chr.16: 3657109-3657109 on Build GRCh38
rs763348516 Chr.16: 3657082-3657082 on Build GRCh38
rs750456471 Chr.16: 3657154-3657154 on Build GRCh38
rs150933932 Chr.16: 3657065-3657065 on Build GRCh38

View Full Product Details