Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82831463-82831730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCD
Entrez Gene ID
6904
Gene Name
tubulin folding cofactor D
Gene Aliases
SSD-1, tfcD
UniGene
Hs.464391
Gene Chromosome Location
Chr.17: 82752048-82943186 on Build GRCh38
Transcripts
NM_005993.4 XM_005256396.4 XM_006722290.2 XM_011523589.1 XM_011523590.1 XM_011523591.1 XM_011523593.1 XM_017024987.1 XM_017024988.1 XM_017024990.1 NM_024702.2

Gene Symbol
ZNF750
Entrez Gene ID
79755
Gene Name
zinc finger protein 750
Gene Aliases
ZFP750
UniGene
Hs.464391
Gene Chromosome Location
Chr.17: 82829434-82840055 on Build GRCh38
Transcripts
NM_005993.4 XM_005256396.4 XM_006722290.2 XM_011523589.1 XM_011523590.1 XM_011523591.1 XM_011523593.1 XM_017024987.1 XM_017024988.1 XM_017024990.1 NM_024702.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4744897 c.727C>T p.P243S Chr.17: 82831728-82831728 on Build GRCh38
4744896 c.728C>T p.P243L Chr.17: 82831727-82831727 on Build GRCh38
6082486 c.750C>T p.H250H Chr.17: 82831705-82831705 on Build GRCh38
7078224 c.907G>A p.D303N Chr.17: 82831548-82831548 on Build GRCh38
4070956 c.946C>T p.P316S Chr.17: 82831509-82831509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143407397 Chr.17: 82831684-82831684 on Build GRCh38
rs182094871 Chr.17: 82831612-82831612 on Build GRCh38
rs753343701 Chr.17: 82831608-82831608 on Build GRCh38
rs760075902 Chr.17: 82831586-82831586 on Build GRCh38
rs370662607 Chr.17: 82831507-82831507 on Build GRCh38
rs35283702 Chr.17: 82831626-82831626 on Build GRCh38
rs747035190 Chr.17: 82831562-82831562 on Build GRCh38
rs150894363 Chr.17: 82831534-82831534 on Build GRCh38
rs141585758 Chr.17: 82831498-82831498 on Build GRCh38
rs202044757 Chr.17: 82831547-82831547 on Build GRCh38
rs149234218 Chr.17: 82831577-82831577 on Build GRCh38
rs963484103 Chr.17: 82831522-82831522 on Build GRCh38
rs375619355 Chr.17: 82831660-82831660 on Build GRCh38
rs117000095 Chr.17: 82831585-82831585 on Build GRCh38
rs779801257 Chr.17: 82831514-82831514 on Build GRCh38
rs1028994504 Chr.17: 82831542-82831542 on Build GRCh38
rs779034115 Chr.17: 82831618-82831618 on Build GRCh38
rs372580838 Chr.17: 82831594-82831594 on Build GRCh38
rs770299355 Chr.17: 82831672-82831672 on Build GRCh38
rs772392011 Chr.17: 82831549-82831549 on Build GRCh38
rs778751897 Chr.17: 82831528-82831528 on Build GRCh38
rs1056834952 Chr.17: 82831616-82831616 on Build GRCh38
rs758289635 Chr.17: 82831627-82831627 on Build GRCh38
rs778171782 Chr.17: 82831554-82831554 on Build GRCh38
rs199951417 Chr.17: 82831520-82831520 on Build GRCh38
rs749599770 Chr.17: 82831675-82831675 on Build GRCh38
rs754934184 Chr.17: 82831519-82831519 on Build GRCh38
rs137910288 Chr.17: 82831591-82831591 on Build GRCh38
rs937600251 Chr.17: 82831671-82831671 on Build GRCh38
rs750092440 Chr.17: 82831593-82831593 on Build GRCh38
rs370475946 Chr.17: 82831704-82831704 on Build GRCh38
rs34566230 Chr.17: 82831548-82831548 on Build GRCh38
rs201166994 Chr.17: 82831621-82831621 on Build GRCh38
rs35653278 Chr.17: 82831592-82831592 on Build GRCh38
rs765631816 Chr.17: 82831488-82831488 on Build GRCh38
rs1003105439 Chr.17: 82831598-82831598 on Build GRCh38
rs898913707 Chr.17: 82831647-82831647 on Build GRCh38
rs570304182 Chr.17: 82831648-82831648 on Build GRCh38
rs192308712 Chr.17: 82831497-82831497 on Build GRCh38
rs528607524 Chr.17: 82831666-82831666 on Build GRCh38
rs202180755 Chr.17: 82831511-82831511 on Build GRCh38
rs778081819 Chr.17: 82831643-82831643 on Build GRCh38

View Full Product Details