Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88427817-88428050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF469
Entrez Gene ID
84627
Gene Name
zinc finger protein 469
Gene Aliases
BCS, BCS1
UniGene
Hs.54925
Gene Chromosome Location
Chr.16: 88424805-88440757 on Build GRCh38
Transcripts
NM_001127464.2 XM_017023784.1 XM_017023785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758556 c.373G>A p.A125T Chr.16: 88427843-88427843 on Build GRCh38
7285413 c.455delC p.Q154fs*192 Chr.16: 88427925-88427925 on Build GRCh38
4407574 c.454A>C p.T152P Chr.16: 88427924-88427924 on Build GRCh38
5461580 c.402C>T p.D134D Chr.16: 88427872-88427872 on Build GRCh38
5542249 c.403G>A p.E135K Chr.16: 88427873-88427873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116028037 Chr.16: 88427851-88427851 on Build GRCh38
rs989169995 Chr.16: 88428026-88428026 on Build GRCh38
rs1046573655 Chr.16: 88427844-88427844 on Build GRCh38
rs569036398 Chr.16: 88427872-88427872 on Build GRCh38
rs545236998 Chr.16: 88427941-88427941 on Build GRCh38
rs781096189 Chr.16: 88427940-88427940 on Build GRCh38
rs769547100 Chr.16: 88427952-88427952 on Build GRCh38
rs771620429 Chr.16: 88428002-88428002 on Build GRCh38
rs761379794 Chr.16: 88427996-88427996 on Build GRCh38
rs757211390 Chr.16: 88427930-88427930 on Build GRCh38
rs773666313 Chr.16: 88427995-88427995 on Build GRCh38
rs530740920 Chr.16: 88427989-88427989 on Build GRCh38
rs936284356 Chr.16: 88428001-88428001 on Build GRCh38
rs889774408 Chr.16: 88427951-88427951 on Build GRCh38
rs765601628 Chr.16: 88427850-88427850 on Build GRCh38
rs565385445 Chr.16: 88427918-88427918 on Build GRCh38
rs994836125 Chr.16: 88427916-88427916 on Build GRCh38
rs951733030 Chr.16: 88427846-88427846 on Build GRCh38
rs940155253 Chr.16: 88427873-88427873 on Build GRCh38
rs914112832 Chr.16: 88428029-88428029 on Build GRCh38
rs760014906 Chr.16: 88428006-88428006 on Build GRCh38
rs71394098 Chr.16: 88427934-88427934 on Build GRCh38
rs755989926 Chr.16: 88427961-88427961 on Build GRCh38
rs1037180222 Chr.16: 88427903-88427903 on Build GRCh38
rs563480285 Chr.16: 88427962-88427962 on Build GRCh38
rs777251198 Chr.16: 88428003-88428003 on Build GRCh38
rs532620482 Chr.16: 88427927-88427927 on Build GRCh38
rs912413955 Chr.16: 88427945-88427945 on Build GRCh38
rs886052389 Chr.16: 88427846-88427846 on Build GRCh38

View Full Product Details