Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29326003-29326242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713878 c.389T>C p.F130S Chr.21: 29326213-29326213 on Build GRCh38
1029783 c.403G>A p.A135T Chr.21: 29326227-29326227 on Build GRCh38
5858481 c.244A>C p.K82Q Chr.21: 29326068-29326068 on Build GRCh38
1184356 c.261A>C p.L87F Chr.21: 29326085-29326085 on Build GRCh38
4482161 c.257C>T p.P86L Chr.21: 29326081-29326081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769099592 Chr.21: 29326038-29326042 on Build GRCh38
rs768809193 Chr.21: 29326134-29326134 on Build GRCh38
rs754935841 Chr.21: 29326220-29326220 on Build GRCh38
rs200941692 Chr.21: 29326186-29326186 on Build GRCh38
rs961975861 Chr.21: 29326171-29326171 on Build GRCh38
rs747065619 Chr.21: 29326121-29326121 on Build GRCh38
rs954180524 Chr.21: 29326035-29326035 on Build GRCh38
rs200422810 Chr.21: 29326091-29326091 on Build GRCh38
rs375336154 Chr.21: 29326036-29326036 on Build GRCh38
rs762029576 Chr.21: 29326156-29326156 on Build GRCh38
rs776883023 Chr.21: 29326147-29326147 on Build GRCh38
rs368753848 Chr.21: 29326130-29326130 on Build GRCh38
rs751446979 Chr.21: 29326215-29326215 on Build GRCh38
rs146449771 Chr.21: 29326165-29326165 on Build GRCh38
rs536072969 Chr.21: 29326133-29326133 on Build GRCh38
rs746275997 Chr.21: 29326042-29326042 on Build GRCh38
rs373065218 Chr.21: 29326167-29326167 on Build GRCh38
rs762726686 Chr.21: 29326179-29326179 on Build GRCh38
rs984389128 Chr.21: 29326065-29326065 on Build GRCh38
rs776178497 Chr.21: 29326105-29326105 on Build GRCh38

View Full Product Details