Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93186333-93186566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM251
Entrez Gene ID
26175
Gene Name
transmembrane protein 251
Gene Aliases
C14orf109
UniGene
Hs.275352
Gene Chromosome Location
Chr.14: 93184951-93187086 on Build GRCh38
Transcripts
NM_001098621.1 NM_015676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477862 c.171C>T p.H57H Chr.14: 93186446-93186446 on Build GRCh38
3370102 c.284A>G p.K95R Chr.14: 93186559-93186559 on Build GRCh38
1135629 c.223C>T p.L75L Chr.14: 93186384-93186384 on Build GRCh38
1135630 c.109C>T p.L37L Chr.14: 93186384-93186384 on Build GRCh38
1246456 c.169C>G p.H57D Chr.14: 93186444-93186444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189524704 Chr.14: 93186442-93186442 on Build GRCh38
rs1013026962 Chr.14: 93186465-93186465 on Build GRCh38
rs772948029 Chr.14: 93186420-93186420 on Build GRCh38
rs897996630 Chr.14: 93186539-93186539 on Build GRCh38
rs182003827 Chr.14: 93186451-93186451 on Build GRCh38
rs745927141 Chr.14: 93186504-93186504 on Build GRCh38
rs1046401911 Chr.14: 93186512-93186512 on Build GRCh38
rs768469355 Chr.14: 93186374-93186374 on Build GRCh38
rs199821940 Chr.14: 93186507-93186507 on Build GRCh38
rs373296773 Chr.14: 93186441-93186441 on Build GRCh38
rs12879694 Chr.14: 93186501-93186501 on Build GRCh38
rs761216249 Chr.14: 93186506-93186506 on Build GRCh38
rs773144041 Chr.14: 93186527-93186527 on Build GRCh38
rs761666767 Chr.14: 93186398-93186398 on Build GRCh38
rs1047647183 Chr.14: 93186404-93186404 on Build GRCh38
rs371140586 Chr.14: 93186456-93186456 on Build GRCh38
rs756254807 Chr.14: 93186505-93186505 on Build GRCh38
rs780147951 Chr.14: 93186372-93186375 on Build GRCh38
rs201974904 Chr.14: 93186520-93186520 on Build GRCh38
rs367611847 Chr.14: 93186540-93186540 on Build GRCh38
rs751717148 Chr.14: 93186482-93186482 on Build GRCh38
rs759143610 Chr.14: 93186449-93186449 on Build GRCh38
rs771951258 Chr.14: 93186522-93186522 on Build GRCh38
rs61741456 Chr.14: 93186491-93186491 on Build GRCh38
rs748116196 Chr.14: 93186509-93186509 on Build GRCh38
rs758715737 Chr.14: 93186457-93186457 on Build GRCh38
rs772039207 Chr.14: 93186412-93186412 on Build GRCh38
rs61744704 Chr.14: 93186389-93186389 on Build GRCh38
rs781139981 Chr.14: 93186360-93186360 on Build GRCh38
rs186551483 Chr.14: 93186461-93186461 on Build GRCh38
rs753300834 Chr.14: 93186447-93186447 on Build GRCh38
rs746614329 Chr.14: 93186530-93186530 on Build GRCh38

View Full Product Details