Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141490329-141490554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141430591-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC3
Entrez Gene ID
5098
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
PC43, PCDH-GAMMA-C3, PCDH2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141476002-141512977 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC4
Entrez Gene ID
56098
Gene Name
protocadherin gamma subfamily C, 4
Gene Aliases
PCDH-GAMMA-C4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141485174-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Gene Symbol
PCDHGC5
Entrez Gene ID
56097
Gene Name
protocadherin gamma subfamily C, 5
Gene Aliases
PCDH-GAMMA-C5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141489241-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032403.2 NM_018928.2 NM_018929.2 NM_032407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
420858 c.1282G>A p.D428N Chr.5: 141490522-141490522 on Build GRCh38
5356702 c.1241G>A p.R414Q Chr.5: 141490481-141490481 on Build GRCh38
6741291 c.1203G>A p.E401E Chr.5: 141490443-141490443 on Build GRCh38
4400659 c.1299C>T p.S433S Chr.5: 141490539-141490539 on Build GRCh38
5402544 c.1314C>T p.L438L Chr.5: 141490554-141490554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768474199 Chr.5: 141490370-141490370 on Build GRCh38
rs2233605 Chr.5: 141490412-141490412 on Build GRCh38
rs756915110 Chr.5: 141490431-141490431 on Build GRCh38
rs764843194 Chr.5: 141490530-141490530 on Build GRCh38
rs779932482 Chr.5: 141490363-141490363 on Build GRCh38
rs191201177 Chr.5: 141490522-141490522 on Build GRCh38
rs771164683 Chr.5: 141490481-141490481 on Build GRCh38
rs370141879 Chr.5: 141490456-141490456 on Build GRCh38
rs746957706 Chr.5: 141490455-141490455 on Build GRCh38
rs560525159 Chr.5: 141490440-141490440 on Build GRCh38
rs761956816 Chr.5: 141490376-141490376 on Build GRCh38
rs760080199 Chr.5: 141490415-141490415 on Build GRCh38
rs779502898 Chr.5: 141490446-141490446 on Build GRCh38
rs781026604 Chr.5: 141490471-141490471 on Build GRCh38
rs768123119 Chr.5: 141490516-141490516 on Build GRCh38
rs967095367 Chr.5: 141490506-141490506 on Build GRCh38
rs112078596 Chr.5: 141490350-141490350 on Build GRCh38
rs201347968 Chr.5: 141490369-141490369 on Build GRCh38
rs757900187 Chr.5: 141490442-141490442 on Build GRCh38
rs770046796 Chr.5: 141490381-141490381 on Build GRCh38
rs765446736 Chr.5: 141490406-141490407 on Build GRCh38
rs2233606 Chr.5: 141490413-141490413 on Build GRCh38
rs767996336 Chr.5: 141490417-141490417 on Build GRCh38
rs774505854 Chr.5: 141490507-141490507 on Build GRCh38
rs769951029 Chr.5: 141490475-141490475 on Build GRCh38
rs773232677 Chr.5: 141490388-141490388 on Build GRCh38
rs142637733 Chr.5: 141490488-141490488 on Build GRCh38
rs886264588 Chr.5: 141490490-141490490 on Build GRCh38
rs369206085 Chr.5: 141490515-141490515 on Build GRCh38
rs564439931 Chr.5: 141490480-141490480 on Build GRCh38
rs146064810 Chr.5: 141490521-141490521 on Build GRCh38
rs749937052 Chr.5: 141490441-141490441 on Build GRCh38
rs753051237 Chr.5: 141490422-141490422 on Build GRCh38
rs1018384314 Chr.5: 141490427-141490427 on Build GRCh38
rs994009107 Chr.5: 141490364-141490364 on Build GRCh38
rs544031284 Chr.5: 141490498-141490498 on Build GRCh38
rs771968534 Chr.5: 141490356-141490356 on Build GRCh38

View Full Product Details