Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50288328-50288555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFRD2
Entrez Gene ID
7866
Gene Name
interferon related developmental regulator 2
Gene Aliases
IFNRP, SKMc15, SM15
UniGene
Hs.315177
Gene Chromosome Location
Chr.3: 50287732-50292595 on Build GRCh38
Transcripts
NM_006764.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2851182 c.1735C>T p.R579C Chr.3: 50288420-50288420 on Build GRCh38
5728610 c.1439A>G p.K480R Chr.3: 50288410-50288410 on Build GRCh38
2851183 c.1724G>A p.C575Y Chr.3: 50288431-50288431 on Build GRCh38
5399645 c.1666C>T p.R556C Chr.3: 50288489-50288489 on Build GRCh38
5829728 c.1651-3_1651-2insC p.? Chr.3: 50288506-50288507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782025532 Chr.3: 50288481-50288481 on Build GRCh38
rs782342919 Chr.3: 50288470-50288470 on Build GRCh38
rs969667858 Chr.3: 50288408-50288408 on Build GRCh38
rs368351951 Chr.3: 50288514-50288514 on Build GRCh38
rs782169802 Chr.3: 50288363-50288363 on Build GRCh38
rs782776242 Chr.3: 50288477-50288477 on Build GRCh38
rs35360115 Chr.3: 50288481-50288482 on Build GRCh38
rs782352389 Chr.3: 50288431-50288431 on Build GRCh38
rs965404682 Chr.3: 50288501-50288501 on Build GRCh38
rs782161246 Chr.3: 50288488-50288488 on Build GRCh38
rs782083933 Chr.3: 50288376-50288376 on Build GRCh38
rs372680234 Chr.3: 50288439-50288439 on Build GRCh38
rs782263446 Chr.3: 50288402-50288402 on Build GRCh38
rs781804984 Chr.3: 50288375-50288375 on Build GRCh38
rs1002747349 Chr.3: 50288424-50288424 on Build GRCh38
rs988246716 Chr.3: 50288482-50288482 on Build GRCh38
rs909070410 Chr.3: 50288530-50288530 on Build GRCh38
rs782423424 Chr.3: 50288446-50288446 on Build GRCh38
rs781861692 Chr.3: 50288391-50288391 on Build GRCh38
rs782202612 Chr.3: 50288426-50288426 on Build GRCh38
rs782670258 Chr.3: 50288400-50288400 on Build GRCh38
rs376948252 Chr.3: 50288444-50288444 on Build GRCh38
rs782691760 Chr.3: 50288365-50288365 on Build GRCh38
rs781803329 Chr.3: 50288492-50288492 on Build GRCh38
rs923142146 Chr.3: 50288512-50288512 on Build GRCh38
rs782080021 Chr.3: 50288378-50288378 on Build GRCh38
rs374530196 Chr.3: 50288508-50288508 on Build GRCh38
rs782456838 Chr.3: 50288507-50288507 on Build GRCh38
rs782119392 Chr.3: 50288475-50288475 on Build GRCh38
rs781924288 Chr.3: 50288448-50288448 on Build GRCh38
rs781792298 Chr.3: 50288517-50288517 on Build GRCh38
rs782186228 Chr.3: 50288522-50288522 on Build GRCh38
rs114263230 Chr.3: 50288534-50288534 on Build GRCh38
rs782693189 Chr.3: 50288491-50288491 on Build GRCh38
rs781852874 Chr.3: 50288509-50288509 on Build GRCh38
rs1013826830 Chr.3: 50288352-50288352 on Build GRCh38
rs782665480 Chr.3: 50288521-50288521 on Build GRCh38
rs371139300 Chr.3: 50288489-50288489 on Build GRCh38
rs782456305 Chr.3: 50288524-50288524 on Build GRCh38
rs116064489 Chr.3: 50288401-50288401 on Build GRCh38
rs370471265 Chr.3: 50288406-50288406 on Build GRCh38
rs782763232 Chr.3: 50288506-50288507 on Build GRCh38
rs782526934 Chr.3: 50288395-50288395 on Build GRCh38
rs782769007 Chr.3: 50288390-50288390 on Build GRCh38
rs782199903 Chr.3: 50288463-50288463 on Build GRCh38

View Full Product Details