Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49102358-49102583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBR1L
Entrez Gene ID
55716
Gene Name
limb development membrane protein 1 like
Gene Aliases
LIMR
UniGene
Hs.272838
Gene Chromosome Location
Chr.12: 49096551-49111535 on Build GRCh38
Transcripts
NM_001300750.1 NM_001300751.1 NM_018113.3 XM_011538563.2 XM_011538564.2 XM_011538565.2 XM_011538566.2 XM_017019626.1 XM_017019627.1 XM_017019628.1 XM_017019629.1 XM_017019630.1 XM_017019631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476608 c.770A>T p.N257I Chr.12: 49102376-49102376 on Build GRCh38
3671090 c.757C>A p.R253S Chr.12: 49102480-49102480 on Build GRCh38
3671091 c.742C>A p.R248S Chr.12: 49102480-49102480 on Build GRCh38
5841124 c.734C>T p.A245V Chr.12: 49102503-49102503 on Build GRCh38
4557125 c.700G>A p.E234K Chr.12: 49102522-49102522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370048172 Chr.12: 49102424-49102424 on Build GRCh38
rs749533576 Chr.12: 49102556-49102556 on Build GRCh38
rs748093057 Chr.12: 49102416-49102416 on Build GRCh38
rs763224820 Chr.12: 49102442-49102442 on Build GRCh38
rs768431277 Chr.12: 49102531-49102531 on Build GRCh38
rs759716403 Chr.12: 49102435-49102435 on Build GRCh38
rs192520010 Chr.12: 49102457-49102457 on Build GRCh38
rs779806755 Chr.12: 49102398-49102398 on Build GRCh38
rs768135798 Chr.12: 49102464-49102464 on Build GRCh38
rs769945633 Chr.12: 49102546-49102546 on Build GRCh38
rs944401331 Chr.12: 49102419-49102419 on Build GRCh38
rs777810712 Chr.12: 49102517-49102517 on Build GRCh38
rs778687267 Chr.12: 49102493-49102493 on Build GRCh38
rs11168858 Chr.12: 49102459-49102459 on Build GRCh38
rs888834109 Chr.12: 49102507-49102507 on Build GRCh38
rs200579973 Chr.12: 49102479-49102479 on Build GRCh38
rs773022106 Chr.12: 49102434-49102434 on Build GRCh38
rs748863768 Chr.12: 49102411-49102411 on Build GRCh38
rs746887360 Chr.12: 49102518-49102518 on Build GRCh38
rs946659716 Chr.12: 49102380-49102380 on Build GRCh38
rs1008612117 Chr.12: 49102538-49102538 on Build GRCh38
rs888015734 Chr.12: 49102542-49102542 on Build GRCh38
rs375836495 Chr.12: 49102449-49102449 on Build GRCh38
rs746124535 Chr.12: 49102491-49102491 on Build GRCh38
rs372587962 Chr.12: 49102441-49102441 on Build GRCh38
rs201154839 Chr.12: 49102550-49102550 on Build GRCh38
rs373373365 Chr.12: 49102413-49102413 on Build GRCh38
rs553538578 Chr.12: 49102480-49102480 on Build GRCh38
rs776032427 Chr.12: 49102544-49102544 on Build GRCh38
rs749880895 Chr.12: 49102387-49102387 on Build GRCh38
rs758112342 Chr.12: 49102514-49102514 on Build GRCh38
rs377556839 Chr.12: 49102415-49102415 on Build GRCh38
rs769735707 Chr.12: 49102439-49102439 on Build GRCh38
rs755567986 Chr.12: 49102393-49102393 on Build GRCh38
rs201580655 Chr.12: 49102468-49102468 on Build GRCh38
rs748006998 Chr.12: 49102495-49102495 on Build GRCh38
rs753512873 Chr.12: 49102488-49102488 on Build GRCh38

View Full Product Details