Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53379058-53379299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5131751 c.86-6T>G p.? Chr.4: 53379067-53379067 on Build GRCh38
6664177 c.131-5G>C p.? Chr.4: 53379220-53379220 on Build GRCh38
4125088 c.105G>A p.R35R Chr.4: 53379244-53379244 on Build GRCh38
6041226 c.134_146del13 p.N46fs*29 Chr.4: 53379228-53379240 on Build GRCh38
3301138 c.109A>G p.S37G Chr.4: 53379096-53379096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370931474 Chr.4: 53379083-53379083 on Build GRCh38
rs866427712 Chr.4: 53379175-53379178 on Build GRCh38
rs370747439 Chr.4: 53379171-53379171 on Build GRCh38
rs963940604 Chr.4: 53379167-53379168 on Build GRCh38
rs770116385 Chr.4: 53379193-53379196 on Build GRCh38
rs367710840 Chr.4: 53379237-53379237 on Build GRCh38
rs368745990 Chr.4: 53379216-53379216 on Build GRCh38
rs779527379 Chr.4: 53379113-53379113 on Build GRCh38
rs751392086 Chr.4: 53379191-53379191 on Build GRCh38
rs772403593 Chr.4: 53379191-53379191 on Build GRCh38
rs750135082 Chr.4: 53379211-53379211 on Build GRCh38
rs587778364 Chr.4: 53379088-53379088 on Build GRCh38
rs748599096 Chr.4: 53379129-53379131 on Build GRCh38
rs767269409 Chr.4: 53379207-53379207 on Build GRCh38
rs749874617 Chr.4: 53379114-53379114 on Build GRCh38
rs748416079 Chr.4: 53379159-53379159 on Build GRCh38
rs749779629 Chr.4: 53379225-53379230 on Build GRCh38
rs768980265 Chr.4: 53379118-53379118 on Build GRCh38
rs771009502 Chr.4: 53379247-53379249 on Build GRCh38
rs749966994 Chr.4: 53379097-53379097 on Build GRCh38
rs780773356 Chr.4: 53379084-53379084 on Build GRCh38
rs35209549 Chr.4: 53379175-53379175 on Build GRCh38
rs761555170 Chr.4: 53379192-53379192 on Build GRCh38
rs776567006 Chr.4: 53379174-53379174 on Build GRCh38
rs755528845 Chr.4: 53379221-53379221 on Build GRCh38
rs763878554 Chr.4: 53379182-53379182 on Build GRCh38
rs747537858 Chr.4: 53379178-53379178 on Build GRCh38
rs564872406 Chr.4: 53379103-53379103 on Build GRCh38
rs868523649 Chr.4: 53379187-53379187 on Build GRCh38
rs773777761 Chr.4: 53379204-53379207 on Build GRCh38
rs371676507 Chr.4: 53379260-53379260 on Build GRCh38
rs778459581 Chr.4: 53379173-53379173 on Build GRCh38
rs199730748 Chr.4: 53379232-53379232 on Build GRCh38
rs375164549 Chr.4: 53379125-53379125 on Build GRCh38
rs372183751 Chr.4: 53379220-53379220 on Build GRCh38
rs772459076 Chr.4: 53379161-53379161 on Build GRCh38
rs773612742 Chr.4: 53379170-53379170 on Build GRCh38
rs779351623 Chr.4: 53379155-53379155 on Build GRCh38

View Full Product Details