Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48482909-48483164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
UniGene
Hs.23170
Gene Chromosome Location
Chr.X: 48476021-48486364 on Build GRCh38
Transcripts
NM_001282157.1 NM_012280.3 NM_177439.2 XM_005272595.1 XM_011543893.2 XM_011543894.2 XM_011543895.1 XM_017029382.1

SNP Detail

SNP ID Chromosome Location
rs374220638 Chr.X: 48482948-48482948 on Build GRCh38
rs781974063 Chr.X: 48482941-48482941 on Build GRCh38
rs201235879 Chr.X: 48482957-48482957 on Build GRCh38
rs782485981 Chr.X: 48483033-48483033 on Build GRCh38
rs781869814 Chr.X: 48483054-48483054 on Build GRCh38
rs200666639 Chr.X: 48483046-48483046 on Build GRCh38
rs186467511 Chr.X: 48483117-48483117 on Build GRCh38
rs375666026 Chr.X: 48483077-48483077 on Build GRCh38
rs781847401 Chr.X: 48482964-48482964 on Build GRCh38
rs782162905 Chr.X: 48482966-48482966 on Build GRCh38
rs941129956 Chr.X: 48483103-48483103 on Build GRCh38
rs781986380 Chr.X: 48482960-48482960 on Build GRCh38
rs201095751 Chr.X: 48483026-48483026 on Build GRCh38
rs782797266 Chr.X: 48482977-48482977 on Build GRCh38
rs782234988 Chr.X: 48483073-48483073 on Build GRCh38
rs7066831 Chr.X: 48483074-48483074 on Build GRCh38

View Full Product Details