Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69415100-69415361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFU1
Entrez Gene ID
27247
Gene Name
NFU1 iron-sulfur cluster scaffold
Gene Aliases
CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU
UniGene
Hs.430439
Gene Chromosome Location
Chr.2: 69396113-69438123 on Build GRCh38
Transcripts
NM_001002755.2 NM_001002756.2 NM_015700.3 NR_045631.1 NR_045632.1 XM_017003808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5091034 c.379G>T p.E127* Chr.2: 69415290-69415290 on Build GRCh38
443130 c.435T>C p.F145F Chr.2: 69415234-69415234 on Build GRCh38
722013 c.473C>T p.S158L Chr.2: 69415196-69415196 on Build GRCh38
3056039 c.370G>T p.E124* Chr.2: 69415299-69415299 on Build GRCh38
5041112 c.413A>G p.Y138C Chr.2: 69415256-69415256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774943458 Chr.2: 69415320-69415320 on Build GRCh38
rs762320530 Chr.2: 69415301-69415301 on Build GRCh38
rs980613284 Chr.2: 69415149-69415149 on Build GRCh38
rs753869603 Chr.2: 69415176-69415176 on Build GRCh38
rs377568651 Chr.2: 69415325-69415325 on Build GRCh38
rs770281955 Chr.2: 69415219-69415219 on Build GRCh38
rs751635934 Chr.2: 69415224-69415225 on Build GRCh38
rs764895827 Chr.2: 69415180-69415180 on Build GRCh38
rs749272908 Chr.2: 69415237-69415237 on Build GRCh38
rs563984185 Chr.2: 69415144-69415144 on Build GRCh38
rs763302368 Chr.2: 69415135-69415135 on Build GRCh38
rs763838516 Chr.2: 69415302-69415302 on Build GRCh38
rs142553805 Chr.2: 69415272-69415272 on Build GRCh38
rs747420560 Chr.2: 69415205-69415205 on Build GRCh38
rs766567121 Chr.2: 69415220-69415221 on Build GRCh38
rs773077576 Chr.2: 69415152-69415152 on Build GRCh38
rs865820240 Chr.2: 69415184-69415184 on Build GRCh38
rs372901537 Chr.2: 69415177-69415177 on Build GRCh38
rs774399970 Chr.2: 69415251-69415251 on Build GRCh38
rs766454379 Chr.2: 69415174-69415174 on Build GRCh38
rs759609855 Chr.2: 69415280-69415280 on Build GRCh38
rs757520389 Chr.2: 69415210-69415210 on Build GRCh38
rs375766930 Chr.2: 69415293-69415293 on Build GRCh38
rs763229821 Chr.2: 69415217-69415217 on Build GRCh38
rs375242060 Chr.2: 69415319-69415319 on Build GRCh38
rs376562786 Chr.2: 69415155-69415155 on Build GRCh38
rs930758906 Chr.2: 69415292-69415292 on Build GRCh38
rs764155316 Chr.2: 69415151-69415151 on Build GRCh38
rs202151809 Chr.2: 69415173-69415173 on Build GRCh38
rs773240399 Chr.2: 69415178-69415179 on Build GRCh38
rs746433132 Chr.2: 69415218-69415218 on Build GRCh38
rs758074085 Chr.2: 69415190-69415190 on Build GRCh38
rs749796927 Chr.2: 69415250-69415250 on Build GRCh38
rs777232071 Chr.2: 69415193-69415193 on Build GRCh38
rs201144794 Chr.2: 69415295-69415295 on Build GRCh38
rs917257642 Chr.2: 69415239-69415239 on Build GRCh38
rs750719069 Chr.2: 69415328-69415328 on Build GRCh38
rs12474866 Chr.2: 69415258-69415258 on Build GRCh38
rs752245896 Chr.2: 69415187-69415187 on Build GRCh38
rs369352430 Chr.2: 69415340-69415340 on Build GRCh38
rs770896590 Chr.2: 69415271-69415271 on Build GRCh38
rs756627984 Chr.2: 69415334-69415334 on Build GRCh38
rs769275443 Chr.2: 69415296-69415296 on Build GRCh38
rs768949114 Chr.2: 69415249-69415249 on Build GRCh38

View Full Product Details