Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21538740-21539009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLLT10
Entrez Gene ID
8028
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Gene Aliases
AF10
UniGene
Hs.30385
Gene Chromosome Location
Chr.10: 21533349-21743630 on Build GRCh38
Transcripts
NM_001195626.1 NM_001195627.1 NM_001195628.1 NM_001195630.1 NM_001324296.1 NM_001324297.1 NM_004641.3 NR_136736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786663 c.221A>C p.Q74P Chr.10: 21538893-21538893 on Build GRCh38
6498800 c.178C>T p.Q60* Chr.10: 21538850-21538850 on Build GRCh38
3806905 c.189T>A p.T63T Chr.10: 21538861-21538861 on Build GRCh38
6498797 c.178C>T p.Q60* Chr.10: 21538850-21538850 on Build GRCh38
6498799 c.178C>T p.Q60* Chr.10: 21538850-21538850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557751007 Chr.10: 21538944-21538944 on Build GRCh38
rs41277382 Chr.10: 21538852-21538852 on Build GRCh38
rs201107641 Chr.10: 21538795-21538795 on Build GRCh38
rs767322244 Chr.10: 21538820-21538820 on Build GRCh38
rs912250452 Chr.10: 21538814-21538814 on Build GRCh38
rs778545162 Chr.10: 21538897-21538897 on Build GRCh38
rs770234454 Chr.10: 21538785-21538785 on Build GRCh38
rs1045568425 Chr.10: 21538933-21538933 on Build GRCh38
rs975219027 Chr.10: 21538866-21538866 on Build GRCh38
rs747608670 Chr.10: 21538927-21538927 on Build GRCh38
rs751089905 Chr.10: 21538920-21538920 on Build GRCh38
rs965399374 Chr.10: 21538824-21538824 on Build GRCh38
rs200231830 Chr.10: 21538934-21538934 on Build GRCh38
rs536845497 Chr.10: 21538796-21538796 on Build GRCh38
rs1055873507 Chr.10: 21538973-21538973 on Build GRCh38
rs756203559 Chr.10: 21538790-21538790 on Build GRCh38
rs750285569 Chr.10: 21538822-21538822 on Build GRCh38
rs937074685 Chr.10: 21538972-21538972 on Build GRCh38
rs775885951 Chr.10: 21538787-21538787 on Build GRCh38
rs1802669 Chr.10: 21538867-21538867 on Build GRCh38
rs775937321 Chr.10: 21538955-21538955 on Build GRCh38
rs746336042 Chr.10: 21538941-21538941 on Build GRCh38
rs17822302 Chr.10: 21538968-21538968 on Build GRCh38
rs761872642 Chr.10: 21538801-21538801 on Build GRCh38
rs935600354 Chr.10: 21538985-21538985 on Build GRCh38
rs982666718 Chr.10: 21538802-21538802 on Build GRCh38
rs763199436 Chr.10: 21538792-21538792 on Build GRCh38
rs766125843 Chr.10: 21538864-21538864 on Build GRCh38
rs371985192 Chr.10: 21538928-21538928 on Build GRCh38

View Full Product Details