Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26033193-26033464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AB
Entrez Gene ID
8335
Gene Name
histone cluster 1, H2ab
Gene Aliases
H2A/m, H2AFM
UniGene
Hs.248174
Gene Chromosome Location
Chr.6: 26033092-26033568 on Build GRCh38
Transcripts
NM_003513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
450999 c.277G>A p.E93K Chr.6: 26033292-26033292 on Build GRCh38
450998 c.364G>C p.E122Q Chr.6: 26033205-26033205 on Build GRCh38
7427419 c.357G>A p.K119K Chr.6: 26033212-26033212 on Build GRCh38
4524054 c.124G>A p.E42K Chr.6: 26033445-26033445 on Build GRCh38
4690405 c.138T>G p.A46A Chr.6: 26033431-26033431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746059666 Chr.6: 26033379-26033379 on Build GRCh38
rs924983962 Chr.6: 26033324-26033324 on Build GRCh38
rs749260550 Chr.6: 26033224-26033224 on Build GRCh38
rs368773475 Chr.6: 26033313-26033313 on Build GRCh38
rs773812280 Chr.6: 26033353-26033353 on Build GRCh38
rs368489925 Chr.6: 26033410-26033410 on Build GRCh38
rs370267838 Chr.6: 26033335-26033335 on Build GRCh38
rs202245482 Chr.6: 26033340-26033340 on Build GRCh38
rs200711320 Chr.6: 26033428-26033428 on Build GRCh38
rs762353384 Chr.6: 26033273-26033273 on Build GRCh38
rs775754511 Chr.6: 26033248-26033248 on Build GRCh38
rs765439690 Chr.6: 26033340-26033342 on Build GRCh38
rs935533492 Chr.6: 26033295-26033295 on Build GRCh38
rs992177371 Chr.6: 26033237-26033237 on Build GRCh38
rs767463907 Chr.6: 26033300-26033300 on Build GRCh38
rs201159814 Chr.6: 26033433-26033433 on Build GRCh38
rs772726939 Chr.6: 26033430-26033430 on Build GRCh38
rs147664123 Chr.6: 26033389-26033389 on Build GRCh38
rs761147784 Chr.6: 26033260-26033260 on Build GRCh38
rs753982270 Chr.6: 26033369-26033369 on Build GRCh38
rs770202067 Chr.6: 26033423-26033423 on Build GRCh38
rs199820035 Chr.6: 26033395-26033395 on Build GRCh38
rs747836793 Chr.6: 26033427-26033427 on Build GRCh38
rs145791004 Chr.6: 26033445-26033445 on Build GRCh38
rs750151736 Chr.6: 26033302-26033302 on Build GRCh38
rs778531979 Chr.6: 26033425-26033425 on Build GRCh38
rs201179274 Chr.6: 26033446-26033446 on Build GRCh38
rs763677828 Chr.6: 26033350-26033350 on Build GRCh38
rs747595349 Chr.6: 26033383-26033383 on Build GRCh38
rs376858886 Chr.6: 26033305-26033305 on Build GRCh38
rs879273097 Chr.6: 26033311-26033311 on Build GRCh38
rs762167197 Chr.6: 26033320-26033320 on Build GRCh38
rs755950046 Chr.6: 26033304-26033304 on Build GRCh38
rs1032127997 Chr.6: 26033433-26033433 on Build GRCh38
rs535683450 Chr.6: 26033432-26033432 on Build GRCh38
rs773675896 Chr.6: 26033253-26033253 on Build GRCh38
rs149461776 Chr.6: 26033242-26033242 on Build GRCh38
rs773874129 Chr.6: 26033332-26033332 on Build GRCh38
rs1026113965 Chr.6: 26033365-26033365 on Build GRCh38
rs200212980 Chr.6: 26033407-26033407 on Build GRCh38
rs766247523 Chr.6: 26033412-26033412 on Build GRCh38
rs751896989 Chr.6: 26033430-26033431 on Build GRCh38
rs759687233 Chr.6: 26033277-26033278 on Build GRCh38
rs545768553 Chr.6: 26033390-26033390 on Build GRCh38
rs144368034 Chr.6: 26033314-26033314 on Build GRCh38
rs779305980 Chr.6: 26033374-26033374 on Build GRCh38
rs764538950 Chr.6: 26033410-26033424 on Build GRCh38
rs145843332 Chr.6: 26033436-26033436 on Build GRCh38
rs765167833 Chr.6: 26033444-26033444 on Build GRCh38
rs199590202 Chr.6: 26033386-26033386 on Build GRCh38
rs550846028 Chr.6: 26033227-26033227 on Build GRCh38
rs752964380 Chr.6: 26033345-26033345 on Build GRCh38
rs758505439 Chr.6: 26033354-26033355 on Build GRCh38
rs1006839948 Chr.6: 26033345-26033345 on Build GRCh38
rs767384212 Chr.6: 26033357-26033357 on Build GRCh38
rs376675696 Chr.6: 26033434-26033434 on Build GRCh38
rs531499669 Chr.6: 26033356-26033356 on Build GRCh38
rs149772014 Chr.6: 26033419-26033419 on Build GRCh38
rs147415971 Chr.6: 26033329-26033329 on Build GRCh38
rs375280077 Chr.6: 26033421-26033421 on Build GRCh38
rs891536312 Chr.6: 26033416-26033416 on Build GRCh38
rs371969893 Chr.6: 26033415-26033415 on Build GRCh38
rs760714072 Chr.6: 26033411-26033411 on Build GRCh38
rs778423313 Chr.6: 26033382-26033382 on Build GRCh38
rs757744885 Chr.6: 26033420-26033420 on Build GRCh38
rs750417482 Chr.6: 26033358-26033358 on Build GRCh38
rs2230655 Chr.6: 26033278-26033278 on Build GRCh38
rs369896892 Chr.6: 26033442-26033442 on Build GRCh38
rs992562989 Chr.6: 26033373-26033373 on Build GRCh38
rs139976301 Chr.6: 26033347-26033347 on Build GRCh38
rs753038811 Chr.6: 26033417-26033417 on Build GRCh38
rs757144771 Chr.6: 26033296-26033296 on Build GRCh38
rs774006125 Chr.6: 26033399-26033399 on Build GRCh38
rs564578704 Chr.6: 26033334-26033334 on Build GRCh38
rs149017049 Chr.6: 26033440-26033440 on Build GRCh38
rs148691705 Chr.6: 26033288-26033288 on Build GRCh38
rs951654689 Chr.6: 26033285-26033285 on Build GRCh38
rs764725053 Chr.6: 26033263-26033263 on Build GRCh38
rs780337319 Chr.6: 26033220-26033220 on Build GRCh38
rs772623470 Chr.6: 26033338-26033338 on Build GRCh38
rs775276035 Chr.6: 26033264-26033264 on Build GRCh38
rs779179729 Chr.6: 26033316-26033316 on Build GRCh38
rs761299529 Chr.6: 26033403-26033403 on Build GRCh38
rs2230654 Chr.6: 26033413-26033413 on Build GRCh38
rs372993129 Chr.6: 26033379-26033379 on Build GRCh38
rs145481087 Chr.6: 26033380-26033380 on Build GRCh38
rs374464701 Chr.6: 26033368-26033368 on Build GRCh38
rs368585509 Chr.6: 26033371-26033371 on Build GRCh38

View Full Product Details