Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203483213-203483450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRELP
Entrez Gene ID
5549
Gene Name
proline and arginine rich end leucine rich repeat protein
Gene Aliases
MST161, MSTP161, SLRR2A
UniGene
Hs.632481
Gene Chromosome Location
Chr.1: 203475755-203491352 on Build GRCh38
Transcripts
NM_002725.3 NM_201348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5820570 c.192A>T p.P64P Chr.1: 203483376-203483376 on Build GRCh38
5735807 c.221C>T p.P74L Chr.1: 203483405-203483405 on Build GRCh38
6755279 c.257C>T p.A86V Chr.1: 203483441-203483441 on Build GRCh38
5936923 c.203C>T p.S68F Chr.1: 203483387-203483387 on Build GRCh38
678066 c.54C>T p.A18A Chr.1: 203483238-203483238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748732293 Chr.1: 203483426-203483427 on Build GRCh38
rs991095688 Chr.1: 203483370-203483370 on Build GRCh38
rs779989216 Chr.1: 203483368-203483368 on Build GRCh38
rs764766335 Chr.1: 203483262-203483263 on Build GRCh38
rs200674304 Chr.1: 203483284-203483284 on Build GRCh38
rs755433661 Chr.1: 203483289-203483290 on Build GRCh38
rs562945346 Chr.1: 203483268-203483268 on Build GRCh38
rs760441009 Chr.1: 203483277-203483277 on Build GRCh38
rs770590815 Chr.1: 203483365-203483365 on Build GRCh38
rs750728307 Chr.1: 203483300-203483300 on Build GRCh38
rs377272061 Chr.1: 203483404-203483404 on Build GRCh38
rs979976433 Chr.1: 203483299-203483299 on Build GRCh38
rs780528706 Chr.1: 203483314-203483314 on Build GRCh38
rs780688119 Chr.1: 203483252-203483252 on Build GRCh38
rs758066744 Chr.1: 203483274-203483275 on Build GRCh38
rs749803099 Chr.1: 203483254-203483254 on Build GRCh38
rs201763034 Chr.1: 203483315-203483315 on Build GRCh38
rs925510362 Chr.1: 203483353-203483353 on Build GRCh38
rs41313926 Chr.1: 203483281-203483281 on Build GRCh38
rs769225317 Chr.1: 203483257-203483257 on Build GRCh38
rs747336935 Chr.1: 203483288-203483292 on Build GRCh38
rs781533652 Chr.1: 203483295-203483297 on Build GRCh38
rs755371433 Chr.1: 203483328-203483328 on Build GRCh38
rs372688100 Chr.1: 203483416-203483416 on Build GRCh38
rs747695089 Chr.1: 203483258-203483258 on Build GRCh38
rs200948130 Chr.1: 203483409-203483409 on Build GRCh38
rs767842916 Chr.1: 203483298-203483298 on Build GRCh38
rs747594584 Chr.1: 203483355-203483355 on Build GRCh38
rs369802878 Chr.1: 203483413-203483413 on Build GRCh38
rs756518365 Chr.1: 203483313-203483313 on Build GRCh38
rs138509179 Chr.1: 203483290-203483290 on Build GRCh38
rs756609920 Chr.1: 203483238-203483238 on Build GRCh38
rs760918634 Chr.1: 203483407-203483407 on Build GRCh38
rs199573544 Chr.1: 203483280-203483280 on Build GRCh38
rs775556165 Chr.1: 203483399-203483399 on Build GRCh38
rs150506217 Chr.1: 203483425-203483425 on Build GRCh38
rs140637051 Chr.1: 203483285-203483285 on Build GRCh38
rs146960081 Chr.1: 203483359-203483359 on Build GRCh38
rs201296195 Chr.1: 203483379-203483379 on Build GRCh38
rs367741970 Chr.1: 203483269-203483269 on Build GRCh38
rs942142713 Chr.1: 203483322-203483322 on Build GRCh38
rs558443908 Chr.1: 203483406-203483406 on Build GRCh38
rs781644459 Chr.1: 203483245-203483245 on Build GRCh38
rs750036894 Chr.1: 203483273-203483286 on Build GRCh38
rs766710985 Chr.1: 203483408-203483408 on Build GRCh38
rs779264906 Chr.1: 203483410-203483410 on Build GRCh38
rs767750327 Chr.1: 203483405-203483405 on Build GRCh38
rs149463952 Chr.1: 203483400-203483400 on Build GRCh38
rs762019841 Chr.1: 203483291-203483291 on Build GRCh38
rs777215133 Chr.1: 203483419-203483419 on Build GRCh38
rs746622926 Chr.1: 203483362-203483362 on Build GRCh38
rs780337843 Chr.1: 203483282-203483305 on Build GRCh38
rs779536648 Chr.1: 203483329-203483329 on Build GRCh38

View Full Product Details