Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103412883-103413150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD11
Entrez Gene ID
22984
Gene Name
programmed cell death 11
Gene Aliases
ALG-4, ALG4, NFBP, RRP5
UniGene
Hs.239499
Gene Chromosome Location
Chr.10: 103396615-103446295 on Build GRCh38
Transcripts
NM_014976.1 XM_005269647.3 XM_011539538.2 XM_011539539.2 XM_011539540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
683482 c.979G>A p.V327M Chr.10: 103413116-103413116 on Build GRCh38
914374 c.1010G>A p.R337Q Chr.10: 103413147-103413147 on Build GRCh38
4011363 c.999C>T p.C333C Chr.10: 103413136-103413136 on Build GRCh38
6742246 c.1013C>G p.T338S Chr.10: 103413150-103413150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753003449 Chr.10: 103413016-103413016 on Build GRCh38
rs1031889580 Chr.10: 103412909-103412909 on Build GRCh38
rs746706707 Chr.10: 103413113-103413113 on Build GRCh38
rs946482142 Chr.10: 103413127-103413127 on Build GRCh38
rs776950492 Chr.10: 103412949-103412949 on Build GRCh38
rs756725691 Chr.10: 103413079-103413079 on Build GRCh38
rs751701817 Chr.10: 103412947-103412947 on Build GRCh38
rs184099532 Chr.10: 103412961-103412961 on Build GRCh38
rs972389839 Chr.10: 103412967-103412967 on Build GRCh38
rs918276633 Chr.10: 103412976-103412976 on Build GRCh38
rs1056343777 Chr.10: 103413055-103413055 on Build GRCh38
rs572907898 Chr.10: 103412919-103412919 on Build GRCh38
rs992805546 Chr.10: 103412929-103412929 on Build GRCh38
rs11591265 Chr.10: 103412987-103412987 on Build GRCh38
rs191624165 Chr.10: 103412927-103412927 on Build GRCh38
rs374816080 Chr.10: 103413096-103413096 on Build GRCh38
rs777225995 Chr.10: 103413105-103413105 on Build GRCh38
rs750933169 Chr.10: 103413076-103413076 on Build GRCh38
rs755404927 Chr.10: 103413100-103413100 on Build GRCh38
rs756674031 Chr.10: 103413125-103413125 on Build GRCh38
rs889470896 Chr.10: 103413121-103413121 on Build GRCh38
rs754310372 Chr.10: 103413098-103413098 on Build GRCh38
rs561007492 Chr.10: 103412916-103412916 on Build GRCh38
rs766844961 Chr.10: 103413086-103413086 on Build GRCh38
rs141115892 Chr.10: 103412952-103412952 on Build GRCh38
rs939871854 Chr.10: 103413023-103413023 on Build GRCh38
rs562691737 Chr.10: 103413054-103413054 on Build GRCh38
rs916499991 Chr.10: 103413082-103413082 on Build GRCh38
rs1036918154 Chr.10: 103413059-103413059 on Build GRCh38
rs767654766 Chr.10: 103413070-103413070 on Build GRCh38
rs970155156 Chr.10: 103412956-103412956 on Build GRCh38
rs756478719 Chr.10: 103413056-103413056 on Build GRCh38

View Full Product Details