Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1471408-1471635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS1
Entrez Gene ID
26173
Gene Name
integrator complex subunit 1
Gene Aliases
INT1, NET28
UniGene
Hs.532188
Gene Chromosome Location
Chr.7: 1470277-1504382 on Build GRCh38
Transcripts
NM_001080453.2 XM_011515260.1 XM_017011959.1 XM_017011960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502789 c.6835C>T p.R2279W Chr.7: 1471603-1471603 on Build GRCh38
4831298 c.6811C>T p.L2271L Chr.7: 1471627-1471627 on Build GRCh38
3262168 c.6864C>T p.F2288F Chr.7: 1471574-1471574 on Build GRCh38
3726499 c.6857G>A p.S2286N Chr.7: 1471581-1471581 on Build GRCh38
4692712 c.6835C>A p.R2279R Chr.7: 1471603-1471603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966803099 Chr.7: 1471426-1471426 on Build GRCh38
rs968144488 Chr.7: 1471437-1471437 on Build GRCh38
rs201387183 Chr.7: 1471603-1471603 on Build GRCh38
rs943059107 Chr.7: 1471519-1471519 on Build GRCh38
rs370249809 Chr.7: 1471538-1471538 on Build GRCh38
rs200142723 Chr.7: 1471581-1471581 on Build GRCh38
rs201675191 Chr.7: 1471578-1471578 on Build GRCh38
rs915836494 Chr.7: 1471455-1471455 on Build GRCh38
rs911573299 Chr.7: 1471490-1471490 on Build GRCh38
rs931055656 Chr.7: 1471553-1471553 on Build GRCh38
rs902569608 Chr.7: 1471585-1471585 on Build GRCh38
rs918974875 Chr.7: 1471482-1471482 on Build GRCh38
rs199528180 Chr.7: 1471572-1471572 on Build GRCh38
rs374746342 Chr.7: 1471577-1471577 on Build GRCh38
rs200948377 Chr.7: 1471580-1471580 on Build GRCh38
rs929042370 Chr.7: 1471483-1471483 on Build GRCh38
rs539402009 Chr.7: 1471517-1471517 on Build GRCh38
rs769798569 Chr.7: 1471560-1471560 on Build GRCh38
rs111720607 Chr.7: 1471569-1471569 on Build GRCh38
rs963203040 Chr.7: 1471466-1471466 on Build GRCh38
rs1003603356 Chr.7: 1471602-1471602 on Build GRCh38
rs777788844 Chr.7: 1471612-1471612 on Build GRCh38
rs117597074 Chr.7: 1471565-1471565 on Build GRCh38
rs766880476 Chr.7: 1471592-1471592 on Build GRCh38
rs755648002 Chr.7: 1471573-1471575 on Build GRCh38
rs745754803 Chr.7: 1471551-1471551 on Build GRCh38
rs768393731 Chr.7: 1471571-1471571 on Build GRCh38
rs550853378 Chr.7: 1471429-1471429 on Build GRCh38
rs754354443 Chr.7: 1471599-1471599 on Build GRCh38
rs112314352 Chr.7: 1471570-1471570 on Build GRCh38
rs371963252 Chr.7: 1471543-1471543 on Build GRCh38
rs1003467274 Chr.7: 1471427-1471427 on Build GRCh38
rs991406402 Chr.7: 1471447-1471447 on Build GRCh38
rs376254974 Chr.7: 1471523-1471523 on Build GRCh38
rs973264770 Chr.7: 1471467-1471467 on Build GRCh38
rs758638183 Chr.7: 1471609-1471609 on Build GRCh38
rs772138075 Chr.7: 1471539-1471539 on Build GRCh38
rs748431961 Chr.7: 1471529-1471529 on Build GRCh38
rs1035031524 Chr.7: 1471438-1471438 on Build GRCh38
rs777840014 Chr.7: 1471550-1471550 on Build GRCh38
rs759199206 Chr.7: 1471484-1471484 on Build GRCh38
rs200312224 Chr.7: 1471600-1471600 on Build GRCh38
rs570857544 Chr.7: 1471446-1471446 on Build GRCh38
rs918172753 Chr.7: 1471489-1471489 on Build GRCh38

View Full Product Details