Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23796949-23797206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALE
Entrez Gene ID
2582
Gene Name
UDP-galactose-4-epimerase
Gene Aliases
SDR1E1
UniGene
Hs.632380
Gene Chromosome Location
Chr.1: 23795599-23800804 on Build GRCh38
Transcripts
NM_000403.3 NM_001008216.1 NM_001127621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
906390 c.603C>A p.P201P Chr.1: 23797073-23797073 on Build GRCh38
906391 c.550C>T p.R184C Chr.1: 23797126-23797126 on Build GRCh38
2157016 c.638C>T p.S213F Chr.1: 23797038-23797038 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541289041 Chr.1: 23796994-23796994 on Build GRCh38
rs980894729 Chr.1: 23796984-23796984 on Build GRCh38
rs758054406 Chr.1: 23796980-23796980 on Build GRCh38
rs746112534 Chr.1: 23797154-23797154 on Build GRCh38
rs766676606 Chr.1: 23796981-23796981 on Build GRCh38
rs980886796 Chr.1: 23797148-23797148 on Build GRCh38
rs778400468 Chr.1: 23797139-23797139 on Build GRCh38
rs755401861 Chr.1: 23796995-23796995 on Build GRCh38
rs140515600 Chr.1: 23797125-23797125 on Build GRCh38
rs533469192 Chr.1: 23797122-23797122 on Build GRCh38
rs762418394 Chr.1: 23797180-23797180 on Build GRCh38
rs771264576 Chr.1: 23797062-23797062 on Build GRCh38
rs769608538 Chr.1: 23797163-23797163 on Build GRCh38
rs773268623 Chr.1: 23797165-23797165 on Build GRCh38
rs369293480 Chr.1: 23797159-23797159 on Build GRCh38
rs552092525 Chr.1: 23797124-23797124 on Build GRCh38
rs753452780 Chr.1: 23797063-23797063 on Build GRCh38
rs968414187 Chr.1: 23797144-23797144 on Build GRCh38
rs992108341 Chr.1: 23797022-23797022 on Build GRCh38
rs151234596 Chr.1: 23797146-23797146 on Build GRCh38
rs762512073 Chr.1: 23797099-23797099 on Build GRCh38
rs377394005 Chr.1: 23796970-23796970 on Build GRCh38
rs200509458 Chr.1: 23796976-23796976 on Build GRCh38
rs753194662 Chr.1: 23797012-23797012 on Build GRCh38
rs772615641 Chr.1: 23797097-23797097 on Build GRCh38
rs993396581 Chr.1: 23796971-23796971 on Build GRCh38
rs756344019 Chr.1: 23797126-23797128 on Build GRCh38
rs1007328278 Chr.1: 23797044-23797044 on Build GRCh38
rs767619045 Chr.1: 23796989-23797008 on Build GRCh38
rs761690949 Chr.1: 23797078-23797078 on Build GRCh38
rs781703014 Chr.1: 23797008-23797008 on Build GRCh38
rs1039473647 Chr.1: 23797178-23797178 on Build GRCh38
rs530956805 Chr.1: 23797145-23797145 on Build GRCh38
rs121908045 Chr.1: 23797128-23797128 on Build GRCh38
rs199904829 Chr.1: 23797074-23797074 on Build GRCh38
rs3204468 Chr.1: 23797137-23797137 on Build GRCh38
rs751909263 Chr.1: 23796992-23796992 on Build GRCh38
rs756177345 Chr.1: 23797034-23797034 on Build GRCh38
rs779252888 Chr.1: 23797152-23797152 on Build GRCh38
rs909389202 Chr.1: 23797151-23797151 on Build GRCh38
rs749469333 Chr.1: 23797043-23797043 on Build GRCh38
rs926679517 Chr.1: 23797011-23797011 on Build GRCh38
rs769284363 Chr.1: 23797092-23797092 on Build GRCh38
rs780856102 Chr.1: 23797157-23797157 on Build GRCh38
rs768619074 Chr.1: 23797072-23797072 on Build GRCh38
rs956978752 Chr.1: 23797132-23797132 on Build GRCh38
rs752852327 Chr.1: 23797009-23797010 on Build GRCh38
rs766017459 Chr.1: 23797100-23797100 on Build GRCh38
rs779415315 Chr.1: 23796996-23796996 on Build GRCh38
rs929558635 Chr.1: 23797075-23797075 on Build GRCh38
rs559853259 Chr.1: 23797117-23797117 on Build GRCh38
rs768460726 Chr.1: 23797155-23797155 on Build GRCh38
rs143924570 Chr.1: 23797065-23797065 on Build GRCh38
rs767796512 Chr.1: 23797126-23797126 on Build GRCh38
rs756590751 Chr.1: 23797138-23797138 on Build GRCh38
rs778029183 Chr.1: 23797170-23797171 on Build GRCh38
rs777717886 Chr.1: 23797037-23797037 on Build GRCh38

View Full Product Details