Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 106053156-106053409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD3
Entrez Gene ID
143879
Gene Name
kelch repeat and BTB domain containing 3
Gene Aliases
BKLHD3
UniGene
Hs.101949
Gene Chromosome Location
Chr.11: 106046801-106077765 on Build GRCh38
Transcripts
NM_152433.3 NM_198439.2 XM_006718767.3 XM_011542618.2 XM_011542619.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7372952 c.1303A>G p.I435V Chr.11: 106053386-106053386 on Build GRCh38
4694116 c.1530A>G p.T510T Chr.11: 106053159-106053159 on Build GRCh38
7372951 c.1400A>T p.D467V Chr.11: 106053289-106053289 on Build GRCh38
3791134 c.1461T>C p.S487S Chr.11: 106053228-106053228 on Build GRCh38
2109567 c.1511C>A p.A504D Chr.11: 106053178-106053178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997981454 Chr.11: 106053206-106053206 on Build GRCh38
rs377014300 Chr.11: 106053341-106053341 on Build GRCh38
rs567227724 Chr.11: 106053231-106053231 on Build GRCh38
rs146866910 Chr.11: 106053250-106053250 on Build GRCh38
rs755284511 Chr.11: 106053190-106053190 on Build GRCh38
rs143991938 Chr.11: 106053237-106053237 on Build GRCh38
rs754907418 Chr.11: 106053181-106053184 on Build GRCh38
rs771314814 Chr.11: 106053217-106053217 on Build GRCh38
rs756383157 Chr.11: 106053355-106053355 on Build GRCh38
rs752991617 Chr.11: 106053348-106053348 on Build GRCh38
rs536438301 Chr.11: 106053207-106053207 on Build GRCh38
rs763013039 Chr.11: 106053314-106053314 on Build GRCh38
rs751585553 Chr.11: 106053189-106053189 on Build GRCh38
rs748316452 Chr.11: 106053195-106053195 on Build GRCh38
rs751453753 Chr.11: 106053336-106053336 on Build GRCh38
rs766212702 Chr.11: 106053326-106053326 on Build GRCh38
rs778326962 Chr.11: 106053380-106053380 on Build GRCh38
rs557063575 Chr.11: 106053267-106053267 on Build GRCh38
rs963679008 Chr.11: 106053372-106053372 on Build GRCh38
rs142289053 Chr.11: 106053220-106053220 on Build GRCh38
rs781581063 Chr.11: 106053193-106053193 on Build GRCh38
rs770314302 Chr.11: 106053196-106053196 on Build GRCh38
rs780999240 Chr.11: 106053197-106053197 on Build GRCh38
rs758136853 Chr.11: 106053368-106053368 on Build GRCh38
rs761401541 Chr.11: 106053246-106053246 on Build GRCh38
rs778235064 Chr.11: 106053204-106053204 on Build GRCh38

View Full Product Details