Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135486481-135486746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R26
Entrez Gene ID
9858
Gene Name
protein phosphatase 1 regulatory subunit 26
Gene Aliases
KIAA0649
UniGene
Hs.533260
Gene Chromosome Location
Chr.9: 135477650-135488894 on Build GRCh38
Transcripts
NM_014811.3 XM_005263411.2 XM_017015355.1 XM_017015356.1 XM_017015357.1 XM_017015358.1 XM_017015359.1 XM_017015360.1 XM_017015361.1 XM_017015362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7385252 c.2160G>T p.K720N Chr.9: 135486670-135486670 on Build GRCh38
372055 c.2124G>A p.K708K Chr.9: 135486634-135486634 on Build GRCh38
1636086 c.2088C>T p.A696A Chr.9: 135486598-135486598 on Build GRCh38
1193724 c.2029A>G p.K677E Chr.9: 135486539-135486539 on Build GRCh38
204733 c.2025C>T p.D675D Chr.9: 135486535-135486535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768215472 Chr.9: 135486631-135486631 on Build GRCh38
rs758345973 Chr.9: 135486552-135486552 on Build GRCh38
rs772399168 Chr.9: 135486638-135486638 on Build GRCh38
rs751550923 Chr.9: 135486556-135486556 on Build GRCh38
rs760852485 Chr.9: 135486528-135486528 on Build GRCh38
rs927259343 Chr.9: 135486536-135486536 on Build GRCh38
rs569173506 Chr.9: 135486721-135486721 on Build GRCh38
rs751497641 Chr.9: 135486667-135486667 on Build GRCh38
rs199557743 Chr.9: 135486501-135486501 on Build GRCh38
rs552032298 Chr.9: 135486690-135486690 on Build GRCh38
rs755779813 Chr.9: 135486692-135486692 on Build GRCh38
rs767899798 Chr.9: 135486526-135486526 on Build GRCh38
rs753928623 Chr.9: 135486548-135486548 on Build GRCh38
rs189673212 Chr.9: 135486660-135486660 on Build GRCh38
rs777796953 Chr.9: 135486554-135486554 on Build GRCh38
rs766448812 Chr.9: 135486535-135486535 on Build GRCh38
rs773603807 Chr.9: 135486644-135486644 on Build GRCh38
rs748686296 Chr.9: 135486628-135486628 on Build GRCh38
rs557552238 Chr.9: 135486719-135486719 on Build GRCh38
rs538809619 Chr.9: 135486718-135486718 on Build GRCh38
rs571803163 Chr.9: 135486707-135486707 on Build GRCh38
rs757308930 Chr.9: 135486676-135486676 on Build GRCh38
rs767390657 Chr.9: 135486689-135486689 on Build GRCh38
rs370079457 Chr.9: 135486517-135486517 on Build GRCh38
rs367901853 Chr.9: 135486693-135486693 on Build GRCh38
rs762330220 Chr.9: 135486634-135486634 on Build GRCh38
rs776928327 Chr.9: 135486658-135486658 on Build GRCh38
rs749263805 Chr.9: 135486697-135486697 on Build GRCh38
rs61731948 Chr.9: 135486655-135486655 on Build GRCh38
rs147777469 Chr.9: 135486504-135486504 on Build GRCh38
rs745512822 Chr.9: 135486616-135486616 on Build GRCh38
rs199977384 Chr.9: 135486652-135486652 on Build GRCh38
rs748999845 Chr.9: 135486699-135486699 on Build GRCh38
rs374959708 Chr.9: 135486726-135486726 on Build GRCh38
rs527358949 Chr.9: 135486661-135486661 on Build GRCh38
rs779666614 Chr.9: 135486624-135486624 on Build GRCh38
rs374786291 Chr.9: 135486527-135486527 on Build GRCh38
rs781157949 Chr.9: 135486586-135486586 on Build GRCh38
rs754435255 Chr.9: 135486703-135486703 on Build GRCh38
rs184703723 Chr.9: 135486580-135486580 on Build GRCh38
rs779464917 Chr.9: 135486682-135486682 on Build GRCh38
rs938008900 Chr.9: 135486571-135486571 on Build GRCh38
rs778443937 Chr.9: 135486706-135486706 on Build GRCh38
rs772629712 Chr.9: 135486709-135486709 on Build GRCh38

View Full Product Details