Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94250275-94250532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEM
Entrez Gene ID
2669
Gene Name
GTP binding protein overexpressed in skeletal muscle
Gene Aliases
KIR
UniGene
Hs.654463
Gene Chromosome Location
Chr.8: 94249249-94262345 on Build GRCh38
Transcripts
NM_005261.3 NM_181702.2 XM_017013315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1458698 c.681C>T p.N227N Chr.8: 94250520-94250520 on Build GRCh38
5647471 c.775A>T p.K259* Chr.8: 94250426-94250426 on Build GRCh38
5746895 c.727C>T p.R243W Chr.8: 94250474-94250474 on Build GRCh38
1700368 c.710G>A p.R237Q Chr.8: 94250491-94250491 on Build GRCh38
1102547 c.780G>A p.E260E Chr.8: 94250421-94250421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755375862 Chr.8: 94250399-94250399 on Build GRCh38
rs770363121 Chr.8: 94250415-94250415 on Build GRCh38
rs141761903 Chr.8: 94250498-94250498 on Build GRCh38
rs867506754 Chr.8: 94250452-94250452 on Build GRCh38
rs748700800 Chr.8: 94250404-94250404 on Build GRCh38
rs749374110 Chr.8: 94250477-94250477 on Build GRCh38
rs138147292 Chr.8: 94250492-94250492 on Build GRCh38
rs759294461 Chr.8: 94250364-94250364 on Build GRCh38
rs372371105 Chr.8: 94250356-94250356 on Build GRCh38
rs141202489 Chr.8: 94250462-94250462 on Build GRCh38
rs530942518 Chr.8: 94250388-94250388 on Build GRCh38
rs771095990 Chr.8: 94250420-94250420 on Build GRCh38
rs190907564 Chr.8: 94250483-94250483 on Build GRCh38
rs761609378 Chr.8: 94250502-94250502 on Build GRCh38
rs774608878 Chr.8: 94250421-94250421 on Build GRCh38
rs778310132 Chr.8: 94250474-94250474 on Build GRCh38
rs955943063 Chr.8: 94250457-94250457 on Build GRCh38
rs995722737 Chr.8: 94250463-94250463 on Build GRCh38
rs770550698 Chr.8: 94250302-94250302 on Build GRCh38
rs771063860 Chr.8: 94250482-94250482 on Build GRCh38
rs138265181 Chr.8: 94250447-94250447 on Build GRCh38
rs568267104 Chr.8: 94250300-94250300 on Build GRCh38
rs777215195 Chr.8: 94250296-94250296 on Build GRCh38
rs200775391 Chr.8: 94250450-94250450 on Build GRCh38
rs766608259 Chr.8: 94250348-94250348 on Build GRCh38
rs375369112 Chr.8: 94250446-94250446 on Build GRCh38
rs767490302 Chr.8: 94250367-94250367 on Build GRCh38
rs537389614 Chr.8: 94250315-94250315 on Build GRCh38
rs753332084 Chr.8: 94250386-94250386 on Build GRCh38
rs940281240 Chr.8: 94250360-94250360 on Build GRCh38
rs149831559 Chr.8: 94250381-94250381 on Build GRCh38
rs746154399 Chr.8: 94250414-94250417 on Build GRCh38
rs145789382 Chr.8: 94250398-94250398 on Build GRCh38
rs775199448 Chr.8: 94250439-94250439 on Build GRCh38
rs773218409 Chr.8: 94250417-94250417 on Build GRCh38
rs747387622 Chr.8: 94250321-94250321 on Build GRCh38
rs752653844 Chr.8: 94250368-94250368 on Build GRCh38
rs368821999 Chr.8: 94250491-94250491 on Build GRCh38
rs769356943 Chr.8: 94250327-94250327 on Build GRCh38
rs201686704 Chr.8: 94250382-94250382 on Build GRCh38
rs773272814 Chr.8: 94250337-94250337 on Build GRCh38
rs770019011 Chr.8: 94250336-94250336 on Build GRCh38
rs142606096 Chr.8: 94250346-94250346 on Build GRCh38
rs748506771 Chr.8: 94250472-94250472 on Build GRCh38
rs148957520 Chr.8: 94250428-94250428 on Build GRCh38
rs776610861 Chr.8: 94250499-94250499 on Build GRCh38
rs200768882 Chr.8: 94250473-94250473 on Build GRCh38
rs749379436 Chr.8: 94250419-94250419 on Build GRCh38
rs777248979 Chr.8: 94250329-94250329 on Build GRCh38
rs750125754 Chr.8: 94250454-94250454 on Build GRCh38
rs921666967 Chr.8: 94250509-94250509 on Build GRCh38
rs778625070 Chr.8: 94250397-94250397 on Build GRCh38
rs557181794 Chr.8: 94250362-94250362 on Build GRCh38
rs759705261 Chr.8: 94250423-94250423 on Build GRCh38
rs551066990 Chr.8: 94250403-94250403 on Build GRCh38
rs781697524 Chr.8: 94250469-94250469 on Build GRCh38
rs760620813 Chr.8: 94250441-94250441 on Build GRCh38
rs746095122 Chr.8: 94250486-94250486 on Build GRCh38
rs764080201 Chr.8: 94250443-94250443 on Build GRCh38
rs762037099 Chr.8: 94250330-94250330 on Build GRCh38

View Full Product Details