Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33528796-33529047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18B
Entrez Gene ID
441459
Gene Name
ankyrin repeat domain 18B
Gene Aliases
bA255A11.3, bA255A11.5
UniGene
Hs.493710
Gene Chromosome Location
Chr.9: 33524403-33578755 on Build GRCh38
Transcripts
NM_001244752.1 XM_011517881.2 XM_011517883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4484185 c.277C>G p.Q93E Chr.9: 33528797-33528797 on Build GRCh38
3952666 c.321+2T>C p.? Chr.9: 33528843-33528843 on Build GRCh38
6367437 c.325G>A p.V109I Chr.9: 33529003-33529003 on Build GRCh38
6315202 c.279G>T p.Q93H Chr.9: 33528799-33528799 on Build GRCh38
5889079 c.347G>T p.C116F Chr.9: 33529025-33529025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541222895 Chr.9: 33528833-33528833 on Build GRCh38
rs751342735 Chr.9: 33528987-33528987 on Build GRCh38
rs142689832 Chr.9: 33528888-33528888 on Build GRCh38
rs2380775 Chr.9: 33529009-33529009 on Build GRCh38
rs975276791 Chr.9: 33529023-33529023 on Build GRCh38
rs767174813 Chr.9: 33529004-33529004 on Build GRCh38
rs776610303 Chr.9: 33528868-33528868 on Build GRCh38
rs771176134 Chr.9: 33528866-33528866 on Build GRCh38
rs967274336 Chr.9: 33528904-33528904 on Build GRCh38
rs779089892 Chr.9: 33528856-33528856 on Build GRCh38
rs138124170 Chr.9: 33528861-33528861 on Build GRCh38
rs368372377 Chr.9: 33528826-33528826 on Build GRCh38
rs775365186 Chr.9: 33528956-33528956 on Build GRCh38
rs556311124 Chr.9: 33528837-33528837 on Build GRCh38
rs759636580 Chr.9: 33528876-33528876 on Build GRCh38
rs778157667 Chr.9: 33528857-33528857 on Build GRCh38
rs780633442 Chr.9: 33528840-33528840 on Build GRCh38
rs576367472 Chr.9: 33528854-33528854 on Build GRCh38
rs755488065 Chr.9: 33528852-33528852 on Build GRCh38
rs761412790 Chr.9: 33528996-33528996 on Build GRCh38
rs762742300 Chr.9: 33528966-33528966 on Build GRCh38
rs749917800 Chr.9: 33528846-33528846 on Build GRCh38
rs1019409848 Chr.9: 33528997-33528997 on Build GRCh38
rs4008878 Chr.9: 33528859-33528860 on Build GRCh38
rs763819806 Chr.9: 33528984-33528984 on Build GRCh38
rs755608154 Chr.9: 33529021-33529021 on Build GRCh38

View Full Product Details