Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142319623-142319894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY4
Entrez Gene ID
81848
Gene Name
sprouty RTK signaling antagonist 4
Gene Aliases
HH17
UniGene
Hs.323308
Gene Chromosome Location
Chr.5: 142310427-142325055 on Build GRCh38
Transcripts
NM_001127496.1 NM_001293289.1 NM_001293290.1 NM_030964.3 XM_011537685.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251228 c.9C>A p.S3R Chr.5: 142319757-142319757 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534900792 Chr.5: 142319714-142319714 on Build GRCh38
rs756181404 Chr.5: 142319756-142319756 on Build GRCh38
rs576424146 Chr.5: 142319843-142319843 on Build GRCh38
rs771836497 Chr.5: 142319696-142319696 on Build GRCh38
rs999361941 Chr.5: 142319670-142319670 on Build GRCh38
rs755435181 Chr.5: 142319743-142319743 on Build GRCh38
rs764683727 Chr.5: 142319712-142319712 on Build GRCh38
rs761337089 Chr.5: 142319709-142319709 on Build GRCh38
rs748038190 Chr.5: 142319814-142319814 on Build GRCh38
rs769288625 Chr.5: 142319815-142319815 on Build GRCh38
rs149416168 Chr.5: 142319834-142319834 on Build GRCh38
rs201103417 Chr.5: 142319778-142319778 on Build GRCh38
rs567326666 Chr.5: 142319790-142319790 on Build GRCh38
rs775248610 Chr.5: 142319697-142319697 on Build GRCh38
rs370447202 Chr.5: 142319695-142319695 on Build GRCh38
rs966904376 Chr.5: 142319668-142319668 on Build GRCh38
rs545800185 Chr.5: 142319748-142319748 on Build GRCh38
rs746780651 Chr.5: 142319779-142319779 on Build GRCh38
rs202113451 Chr.5: 142319753-142319753 on Build GRCh38
rs200816826 Chr.5: 142319694-142319694 on Build GRCh38
rs761784825 Chr.5: 142319846-142319846 on Build GRCh38
rs200241261 Chr.5: 142319773-142319773 on Build GRCh38
rs895785677 Chr.5: 142319862-142319862 on Build GRCh38
rs776511089 Chr.5: 142319793-142319793 on Build GRCh38
rs375095885 Chr.5: 142319705-142319705 on Build GRCh38
rs368059191 Chr.5: 142319826-142319826 on Build GRCh38
rs372707899 Chr.5: 142319741-142319741 on Build GRCh38
rs766194857 Chr.5: 142319740-142319740 on Build GRCh38
rs754103857 Chr.5: 142319726-142319729 on Build GRCh38
rs76855889 Chr.5: 142319751-142319751 on Build GRCh38
rs538912604 Chr.5: 142319693-142319693 on Build GRCh38
rs777722207 Chr.5: 142319759-142319759 on Build GRCh38
rs1055340887 Chr.5: 142319825-142319825 on Build GRCh38
rs762527197 Chr.5: 142319732-142319732 on Build GRCh38
rs73292478 Chr.5: 142319827-142319827 on Build GRCh38
rs559693040 Chr.5: 142319703-142319703 on Build GRCh38
rs749440600 Chr.5: 142319762-142319762 on Build GRCh38
rs930255647 Chr.5: 142319808-142319808 on Build GRCh38
rs768118947 Chr.5: 142319749-142319749 on Build GRCh38

View Full Product Details