Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30067021-30067282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
UniGene
Hs.513490
Gene Chromosome Location
Chr.16: 30053090-30070420 on Build GRCh38
Transcripts
NM_000034.3 NM_001127617.2 NM_001243177.1 NM_184041.2 NM_184043.2 XM_011545768.2

SNP Detail

SNP ID Chromosome Location
rs768714573 Chr.16: 30067062-30067062 on Build GRCh38
rs1024113434 Chr.16: 30067104-30067104 on Build GRCh38
rs747888913 Chr.16: 30067184-30067184 on Build GRCh38
rs770516115 Chr.16: 30067208-30067208 on Build GRCh38
rs750976538 Chr.16: 30067229-30067229 on Build GRCh38
rs535217608 Chr.16: 30067053-30067053 on Build GRCh38
rs546976628 Chr.16: 30067072-30067072 on Build GRCh38
rs9783783 Chr.16: 30067171-30067171 on Build GRCh38
rs933445777 Chr.16: 30067165-30067165 on Build GRCh38
rs769709094 Chr.16: 30067194-30067194 on Build GRCh38
rs773340964 Chr.16: 30067198-30067198 on Build GRCh38
rs879225228 Chr.16: 30067228-30067228 on Build GRCh38
rs1033625708 Chr.16: 30067213-30067213 on Build GRCh38
rs371602498 Chr.16: 30067249-30067249 on Build GRCh38
rs775119400 Chr.16: 30067223-30067223 on Build GRCh38
rs900204785 Chr.16: 30067127-30067127 on Build GRCh38
rs770061959 Chr.16: 30067076-30067079 on Build GRCh38
rs34209145 Chr.16: 30067179-30067179 on Build GRCh38
rs762219882 Chr.16: 30067224-30067224 on Build GRCh38
rs910941761 Chr.16: 30067182-30067182 on Build GRCh38
rs76642620 Chr.16: 30067126-30067126 on Build GRCh38
rs772798937 Chr.16: 30067215-30067216 on Build GRCh38
rs756024464 Chr.16: 30067253-30067253 on Build GRCh38
rs201718271 Chr.16: 30067252-30067252 on Build GRCh38
rs781628911 Chr.16: 30067247-30067247 on Build GRCh38
rs923451636 Chr.16: 30067131-30067131 on Build GRCh38
rs959386198 Chr.16: 30067214-30067214 on Build GRCh38
rs565677087 Chr.16: 30067116-30067116 on Build GRCh38
rs879002625 Chr.16: 30067229-30067229 on Build GRCh38
rs903764420 Chr.16: 30067060-30067060 on Build GRCh38
rs759238234 Chr.16: 30067215-30067215 on Build GRCh38
rs555628582 Chr.16: 30067234-30067234 on Build GRCh38
rs773684656 Chr.16: 30067209-30067209 on Build GRCh38
rs774875494 Chr.16: 30067206-30067206 on Build GRCh38
rs944622649 Chr.16: 30067114-30067114 on Build GRCh38
rs759561783 Chr.16: 30067227-30067227 on Build GRCh38
rs778867595 Chr.16: 30067089-30067089 on Build GRCh38
rs751539995 Chr.16: 30067244-30067244 on Build GRCh38
rs762779941 Chr.16: 30067200-30067200 on Build GRCh38
rs772305686 Chr.16: 30067176-30067176 on Build GRCh38
rs373517060 Chr.16: 30067256-30067256 on Build GRCh38
rs982361771 Chr.16: 30067212-30067212 on Build GRCh38
rs1021550064 Chr.16: 30067183-30067183 on Build GRCh38
rs368691859 Chr.16: 30067243-30067243 on Build GRCh38
rs1013644650 Chr.16: 30067094-30067094 on Build GRCh38
rs541579725 Chr.16: 30067260-30067260 on Build GRCh38
rs35257049 Chr.16: 30067094-30067095 on Build GRCh38
rs530878836 Chr.16: 30067073-30067073 on Build GRCh38
rs755235804 Chr.16: 30067245-30067245 on Build GRCh38
rs1009712069 Chr.16: 30067174-30067174 on Build GRCh38
rs868546910 Chr.16: 30067246-30067247 on Build GRCh38
rs999934496 Chr.16: 30067130-30067130 on Build GRCh38
rs779963072 Chr.16: 30067047-30067047 on Build GRCh38
rs376849543 Chr.16: 30067217-30067217 on Build GRCh38
rs969444594 Chr.16: 30067113-30067113 on Build GRCh38
rs576122083 Chr.16: 30067151-30067151 on Build GRCh38
rs985178439 Chr.16: 30067122-30067122 on Build GRCh38
rs766640275 Chr.16: 30067153-30067153 on Build GRCh38

View Full Product Details