Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10631415-10631684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A2
Entrez Gene ID
57153
Gene Name
solute carrier family 44 member 2
Gene Aliases
CTL2, PP1292
UniGene
Hs.534560
Gene Chromosome Location
Chr.19: 10602445-10644559 on Build GRCh38
Transcripts
NM_001145056.1 NM_020428.3 XM_005259997.1 XM_005259999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234101 c.502+3G>A p.? Chr.19: 10631538-10631538 on Build GRCh38
6353694 c.455A>G p.Q152R Chr.19: 10631494-10631494 on Build GRCh38
6406886 c.533C>A p.A178D Chr.19: 10631656-10631656 on Build GRCh38
6782317 c.516C>T p.P172P Chr.19: 10631645-10631645 on Build GRCh38
7095524 c.451G>A p.E151K Chr.19: 10631484-10631484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892936015 Chr.19: 10631516-10631516 on Build GRCh38
rs374051348 Chr.19: 10631603-10631603 on Build GRCh38
rs981410958 Chr.19: 10631477-10631477 on Build GRCh38
rs745789435 Chr.19: 10631627-10631627 on Build GRCh38
rs777795867 Chr.19: 10631608-10631608 on Build GRCh38
rs953396456 Chr.19: 10631660-10631660 on Build GRCh38
rs752370251 Chr.19: 10631438-10631438 on Build GRCh38
rs377482215 Chr.19: 10631533-10631533 on Build GRCh38
rs748376597 Chr.19: 10631484-10631484 on Build GRCh38
rs758794792 Chr.19: 10631465-10631465 on Build GRCh38
rs755525920 Chr.19: 10631600-10631600 on Build GRCh38
rs768234084 Chr.19: 10631649-10631649 on Build GRCh38
rs147820753 Chr.19: 10631490-10631490 on Build GRCh38
rs377075783 Chr.19: 10631632-10631632 on Build GRCh38
rs1029939771 Chr.19: 10631575-10631575 on Build GRCh38
rs781288207 Chr.19: 10631476-10631476 on Build GRCh38
rs1054210036 Chr.19: 10631502-10631502 on Build GRCh38
rs756220054 Chr.19: 10631605-10631605 on Build GRCh38
rs201605858 Chr.19: 10631472-10631472 on Build GRCh38
rs1560711 Chr.19: 10631611-10631611 on Build GRCh38
rs372502995 Chr.19: 10631545-10631545 on Build GRCh38
rs767835027 Chr.19: 10631549-10631550 on Build GRCh38
rs2288904 Chr.19: 10631494-10631494 on Build GRCh38
rs769965009 Chr.19: 10631487-10631487 on Build GRCh38
rs753542585 Chr.19: 10631568-10631568 on Build GRCh38
rs1039168030 Chr.19: 10631546-10631546 on Build GRCh38
rs781233716 Chr.19: 10631602-10631602 on Build GRCh38
rs962833193 Chr.19: 10631626-10631626 on Build GRCh38
rs949113440 Chr.19: 10631539-10631539 on Build GRCh38
rs755773906 Chr.19: 10631441-10631441 on Build GRCh38
rs915916399 Chr.19: 10631463-10631463 on Build GRCh38
rs373190377 Chr.19: 10631645-10631645 on Build GRCh38
rs971446075 Chr.19: 10631631-10631631 on Build GRCh38
rs369733388 Chr.19: 10631452-10631452 on Build GRCh38
rs760046078 Chr.19: 10631646-10631646 on Build GRCh38
rs558954316 Chr.19: 10631654-10631654 on Build GRCh38
rs149430874 Chr.19: 10631655-10631655 on Build GRCh38
rs997580548 Chr.19: 10631564-10631564 on Build GRCh38
rs570362442 Chr.19: 10631610-10631610 on Build GRCh38
rs778903653 Chr.19: 10631622-10631622 on Build GRCh38
rs755009235 Chr.19: 10631475-10631475 on Build GRCh38
rs369618308 Chr.19: 10631571-10631571 on Build GRCh38
rs775200654 Chr.19: 10631531-10631531 on Build GRCh38
rs34814228 Chr.19: 10631555-10631556 on Build GRCh38
rs767116646 Chr.19: 10631507-10631507 on Build GRCh38
rs748964300 Chr.19: 10631493-10631493 on Build GRCh38
rs924564833 Chr.19: 10631633-10631633 on Build GRCh38
rs953342774 Chr.19: 10631437-10631437 on Build GRCh38
rs752028664 Chr.19: 10631584-10631584 on Build GRCh38
rs780481027 Chr.19: 10631470-10631470 on Build GRCh38
rs766760652 Chr.19: 10631572-10631572 on Build GRCh38
rs901520261 Chr.19: 10631547-10631547 on Build GRCh38
rs750820221 Chr.19: 10631464-10631464 on Build GRCh38
rs577326622 Chr.19: 10631656-10631656 on Build GRCh38
rs867102703 Chr.19: 10631569-10631569 on Build GRCh38
rs373481388 Chr.19: 10631504-10631504 on Build GRCh38

View Full Product Details