Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90960285-90960544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCCD1
Entrez Gene ID
91433
Gene Name
RCC1 domain containing 1
Gene Aliases
-
UniGene
Hs.655895
Gene Chromosome Location
Chr.15: 90954864-90964858 on Build GRCh38
Transcripts
NM_001017919.1 NM_033544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
355654 c.868T>G p.F290V Chr.15: 90960417-90960417 on Build GRCh38
4567932 c.929_930GG>AA p.R310Q Chr.15: 90960478-90960479 on Build GRCh38
2140570 c.850T>C p.F284L Chr.15: 90960399-90960399 on Build GRCh38
4578628 c.928C>T p.R310W Chr.15: 90960477-90960477 on Build GRCh38
1240027 c.834T>C p.D278D Chr.15: 90960383-90960383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771424433 Chr.15: 90960421-90960421 on Build GRCh38
rs773642630 Chr.15: 90960323-90960323 on Build GRCh38
rs748322026 Chr.15: 90960379-90960379 on Build GRCh38
rs778133221 Chr.15: 90960408-90960408 on Build GRCh38
rs376275450 Chr.15: 90960318-90960318 on Build GRCh38
rs562663393 Chr.15: 90960307-90960307 on Build GRCh38
rs370018813 Chr.15: 90960492-90960492 on Build GRCh38
rs759029457 Chr.15: 90960438-90960438 on Build GRCh38
rs527421305 Chr.15: 90960477-90960477 on Build GRCh38
rs767842428 Chr.15: 90960522-90960522 on Build GRCh38
rs748529590 Chr.15: 90960320-90960320 on Build GRCh38
rs770377968 Chr.15: 90960328-90960328 on Build GRCh38
rs142701764 Chr.15: 90960394-90960394 on Build GRCh38
rs779137159 Chr.15: 90960376-90960376 on Build GRCh38
rs776177411 Chr.15: 90960333-90960333 on Build GRCh38
rs751495755 Chr.15: 90960354-90960354 on Build GRCh38
rs867078334 Chr.15: 90960409-90960409 on Build GRCh38
rs762721279 Chr.15: 90960453-90960453 on Build GRCh38
rs960968235 Chr.15: 90960332-90960332 on Build GRCh38
rs772386570 Chr.15: 90960396-90960396 on Build GRCh38
rs374753986 Chr.15: 90960341-90960341 on Build GRCh38
rs755068763 Chr.15: 90960375-90960375 on Build GRCh38
rs757241432 Chr.15: 90960367-90960367 on Build GRCh38
rs140875352 Chr.15: 90960322-90960322 on Build GRCh38
rs761199575 Chr.15: 90960325-90960325 on Build GRCh38
rs773449208 Chr.15: 90960464-90960483 on Build GRCh38
rs545375745 Chr.15: 90960484-90960484 on Build GRCh38
rs762746677 Chr.15: 90960347-90960347 on Build GRCh38
rs373221197 Chr.15: 90960315-90960315 on Build GRCh38
rs746912439 Chr.15: 90960393-90960394 on Build GRCh38
rs774633417 Chr.15: 90960498-90960498 on Build GRCh38
rs150144745 Chr.15: 90960442-90960442 on Build GRCh38
rs973623062 Chr.15: 90960519-90960519 on Build GRCh38
rs923179947 Chr.15: 90960397-90960397 on Build GRCh38
rs777064524 Chr.15: 90960422-90960422 on Build GRCh38
rs147224053 Chr.15: 90960368-90960368 on Build GRCh38
rs769195154 Chr.15: 90960440-90960440 on Build GRCh38
rs767546137 Chr.15: 90960465-90960465 on Build GRCh38
rs768676476 Chr.15: 90960448-90960449 on Build GRCh38
rs141767992 Chr.15: 90960349-90960349 on Build GRCh38
rs1867220 Chr.15: 90960522-90960522 on Build GRCh38
rs764929862 Chr.15: 90960337-90960337 on Build GRCh38
rs780191249 Chr.15: 90960371-90960371 on Build GRCh38
rs752915078 Chr.15: 90960499-90960499 on Build GRCh38
rs992609316 Chr.15: 90960399-90960399 on Build GRCh38
rs778888317 Chr.15: 90960498-90960498 on Build GRCh38
rs1023138771 Chr.15: 90960316-90960316 on Build GRCh38
rs367698055 Chr.15: 90960505-90960505 on Build GRCh38
rs747415618 Chr.15: 90960411-90960411 on Build GRCh38
rs61742976 Chr.15: 90960454-90960454 on Build GRCh38
rs763035264 Chr.15: 90960463-90960464 on Build GRCh38
rs771895519 Chr.15: 90960310-90960312 on Build GRCh38
rs909516689 Chr.15: 90960513-90960513 on Build GRCh38
rs202059759 Chr.15: 90960523-90960523 on Build GRCh38
rs548495284 Chr.15: 90960342-90960342 on Build GRCh38
rs771065368 Chr.15: 90960490-90960490 on Build GRCh38
rs964561311 Chr.15: 90960369-90960369 on Build GRCh38
rs759878742 Chr.15: 90960503-90960504 on Build GRCh38
rs778037341 Chr.15: 90960514-90960514 on Build GRCh38
rs761762986 Chr.15: 90960463-90960463 on Build GRCh38

View Full Product Details