Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70372936-70373189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNJ2BP
Entrez Gene ID
55333
Gene Name
synaptojanin 2 binding protein
Gene Aliases
ARIP2, OMP25
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70366496-70417090 on Build GRCh38
Transcripts
NM_018373.2 NM_001202547.2 NM_001202548.2 NM_001202549.2

Gene Symbol
SYNJ2BP-COX16
Entrez Gene ID
100529257
Gene Name
SYNJ2BP-COX16 readthrough
Gene Aliases
-
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70325081-70417090 on Build GRCh38
Transcripts
NM_018373.2 NM_001202547.2 NM_001202548.2 NM_001202549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3401441 c.321A>G p.I107M Chr.14: 70373108-70373108 on Build GRCh38
957371 c.329G>A p.R110Q Chr.14: 70373100-70373100 on Build GRCh38
957370 c.331G>T p.G111C Chr.14: 70373098-70373098 on Build GRCh38
295684 c.377C>T p.P126L Chr.14: 70373052-70373052 on Build GRCh38
7187192 c.417G>A p.M139I Chr.14: 70373012-70373012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991049580 Chr.14: 70373118-70373118 on Build GRCh38
rs924282007 Chr.14: 70372962-70372963 on Build GRCh38
rs992967152 Chr.14: 70372966-70372966 on Build GRCh38
rs970686971 Chr.14: 70373133-70373133 on Build GRCh38
rs74324732 Chr.14: 70372986-70372986 on Build GRCh38
rs45565842 Chr.14: 70373018-70373018 on Build GRCh38
rs200172973 Chr.14: 70372962-70372962 on Build GRCh38
rs897145913 Chr.14: 70373072-70373072 on Build GRCh38
rs777704021 Chr.14: 70373088-70373089 on Build GRCh38
rs774963810 Chr.14: 70372978-70372978 on Build GRCh38
rs780495617 Chr.14: 70372967-70372967 on Build GRCh38
rs758972278 Chr.14: 70373001-70373001 on Build GRCh38
rs780404095 Chr.14: 70373097-70373097 on Build GRCh38
rs369169483 Chr.14: 70373005-70373005 on Build GRCh38
rs373971318 Chr.14: 70373079-70373079 on Build GRCh38
rs138860992 Chr.14: 70373004-70373004 on Build GRCh38
rs765442650 Chr.14: 70372963-70372963 on Build GRCh38
rs749558267 Chr.14: 70373099-70373099 on Build GRCh38
rs776148557 Chr.14: 70372990-70372990 on Build GRCh38
rs369607687 Chr.14: 70373144-70373144 on Build GRCh38
rs1050340767 Chr.14: 70372998-70372998 on Build GRCh38
rs757162113 Chr.14: 70373042-70373042 on Build GRCh38
rs376970622 Chr.14: 70373109-70373109 on Build GRCh38
rs376670851 Chr.14: 70372982-70372982 on Build GRCh38
rs759072342 Chr.14: 70373158-70373158 on Build GRCh38
rs200225131 Chr.14: 70373149-70373149 on Build GRCh38
rs958489560 Chr.14: 70373117-70373117 on Build GRCh38
rs867663901 Chr.14: 70373100-70373100 on Build GRCh38
rs748471527 Chr.14: 70373120-70373120 on Build GRCh38
rs769994749 Chr.14: 70372965-70372965 on Build GRCh38
rs779369416 Chr.14: 70373110-70373110 on Build GRCh38
rs142365285 Chr.14: 70373130-70373130 on Build GRCh38
rs762705588 Chr.14: 70373015-70373015 on Build GRCh38
rs749232433 Chr.14: 70373131-70373132 on Build GRCh38
rs780999249 Chr.14: 70373062-70373062 on Build GRCh38
rs1053488 Chr.14: 70372992-70372992 on Build GRCh38
rs202186250 Chr.14: 70372972-70372972 on Build GRCh38
rs749661485 Chr.14: 70372970-70372970 on Build GRCh38
rs770695069 Chr.14: 70373145-70373145 on Build GRCh38
rs1022475615 Chr.14: 70373150-70373150 on Build GRCh38
rs191854190 Chr.14: 70373160-70373160 on Build GRCh38
rs530067652 Chr.14: 70373031-70373031 on Build GRCh38
rs772561688 Chr.14: 70373121-70373121 on Build GRCh38
rs548275339 Chr.14: 70373087-70373087 on Build GRCh38

View Full Product Details