Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82032240-82032513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAC3
Entrez Gene ID
5881
Gene Name
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
Gene Aliases
-
UniGene
Hs.45002
Gene Chromosome Location
Chr.17: 82031656-82034204 on Build GRCh38
Transcripts
NM_001316307.1 NM_005052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
388501 c.87C>T p.P29P Chr.17: 82032438-82032438 on Build GRCh38
7152893 c.48G>C p.K16N Chr.17: 82032399-82032399 on Build GRCh38
5562098 c.87C>G p.P29P Chr.17: 82032438-82032438 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004821935 Chr.17: 82032287-82032287 on Build GRCh38
rs528509306 Chr.17: 82032369-82032369 on Build GRCh38
rs373875259 Chr.17: 82032375-82032375 on Build GRCh38
rs1030252632 Chr.17: 82032470-82032470 on Build GRCh38
rs868457310 Chr.17: 82032288-82032288 on Build GRCh38
rs956945071 Chr.17: 82032469-82032469 on Build GRCh38
rs774018137 Chr.17: 82032493-82032493 on Build GRCh38
rs774983679 Chr.17: 82032390-82032390 on Build GRCh38
rs750388697 Chr.17: 82032427-82032427 on Build GRCh38
rs976384899 Chr.17: 82032271-82032271 on Build GRCh38
rs953239030 Chr.17: 82032495-82032495 on Build GRCh38
rs561408315 Chr.17: 82032362-82032362 on Build GRCh38
rs141164336 Chr.17: 82032450-82032450 on Build GRCh38
rs985835274 Chr.17: 82032306-82032306 on Build GRCh38
rs748463837 Chr.17: 82032456-82032456 on Build GRCh38
rs761553011 Chr.17: 82032496-82032496 on Build GRCh38
rs375609101 Chr.17: 82032384-82032384 on Build GRCh38
rs542731759 Chr.17: 82032357-82032357 on Build GRCh38
rs369866684 Chr.17: 82032391-82032391 on Build GRCh38
rs182602051 Chr.17: 82032266-82032266 on Build GRCh38
rs769174564 Chr.17: 82032482-82032482 on Build GRCh38
rs778394723 Chr.17: 82032477-82032477 on Build GRCh38
rs377327873 Chr.17: 82032335-82032335 on Build GRCh38
rs756151814 Chr.17: 82032428-82032428 on Build GRCh38
rs768301118 Chr.17: 82032492-82032492 on Build GRCh38
rs888459736 Chr.17: 82032399-82032399 on Build GRCh38
rs931626117 Chr.17: 82032305-82032305 on Build GRCh38
rs772597902 Chr.17: 82032461-82032461 on Build GRCh38
rs769407343 Chr.17: 82032355-82032355 on Build GRCh38
rs756206765 Chr.17: 82032352-82032352 on Build GRCh38
rs749561411 Chr.17: 82032358-82032358 on Build GRCh38
rs1004679834 Chr.17: 82032402-82032402 on Build GRCh38
rs923211083 Chr.17: 82032292-82032292 on Build GRCh38
rs565087961 Chr.17: 82032486-82032486 on Build GRCh38
rs1015142391 Chr.17: 82032259-82032259 on Build GRCh38
rs748755776 Chr.17: 82032489-82032489 on Build GRCh38
rs370032504 Chr.17: 82032360-82032360 on Build GRCh38
rs762728412 Chr.17: 82032411-82032411 on Build GRCh38
rs1026908105 Chr.17: 82032381-82032381 on Build GRCh38
rs936373005 Chr.17: 82032344-82032344 on Build GRCh38
rs747753571 Chr.17: 82032343-82032344 on Build GRCh38
rs780227170 Chr.17: 82032430-82032430 on Build GRCh38
rs781114791 Chr.17: 82032341-82032341 on Build GRCh38
rs753992755 Chr.17: 82032439-82032439 on Build GRCh38
rs750447889 Chr.17: 82032345-82032345 on Build GRCh38
rs763796246 Chr.17: 82032420-82032420 on Build GRCh38
rs371297819 Chr.17: 82032480-82032480 on Build GRCh38
rs1047073470 Chr.17: 82032373-82032373 on Build GRCh38
rs772638672 Chr.17: 82032370-82032370 on Build GRCh38
rs1018963209 Chr.17: 82032460-82032460 on Build GRCh38
rs767390414 Chr.17: 82032426-82032426 on Build GRCh38
rs540611153 Chr.17: 82032377-82032377 on Build GRCh38
rs147951537 Chr.17: 82032423-82032423 on Build GRCh38

View Full Product Details