Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127435823-127436050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF79
Entrez Gene ID
7633
Gene Name
zinc finger protein 79
Gene Aliases
pT7
UniGene
Hs.522399
Gene Chromosome Location
Chr.9: 127424374-127446289 on Build GRCh38
Transcripts
NM_001286696.1 NM_001286697.1 NM_001286698.1 NM_001322260.1 NM_007135.2 XM_006717279.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5012080 c.247C>A p.Q83K Chr.9: 127435922-127435922 on Build GRCh38
6114637 c.237T>G p.L79L Chr.9: 127435912-127435912 on Build GRCh38
118017 c.313C>T p.R105* Chr.9: 127435988-127435988 on Build GRCh38
3654419 c.245C>T p.S82F Chr.9: 127435920-127435920 on Build GRCh38
238503 c.295C>T p.L99L Chr.9: 127435970-127435970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200725787 Chr.9: 127435996-127435996 on Build GRCh38
rs766356170 Chr.9: 127435973-127435973 on Build GRCh38
rs924935459 Chr.9: 127436011-127436011 on Build GRCh38
rs564502090 Chr.9: 127435879-127435879 on Build GRCh38
rs760732822 Chr.9: 127435971-127435971 on Build GRCh38
rs145981250 Chr.9: 127435913-127435913 on Build GRCh38
rs752942190 Chr.9: 127435988-127435988 on Build GRCh38
rs761658837 Chr.9: 127435963-127435963 on Build GRCh38
rs772964724 Chr.9: 127435970-127435970 on Build GRCh38
rs778208151 Chr.9: 127435992-127435992 on Build GRCh38
rs148235940 Chr.9: 127435978-127435978 on Build GRCh38
rs774096726 Chr.9: 127436026-127436026 on Build GRCh38
rs771760163 Chr.9: 127436031-127436031 on Build GRCh38
rs779229429 Chr.9: 127435905-127435905 on Build GRCh38
rs754363719 Chr.9: 127435932-127435932 on Build GRCh38
rs149728551 Chr.9: 127435974-127435974 on Build GRCh38
rs757843744 Chr.9: 127435979-127435979 on Build GRCh38
rs762741047 Chr.9: 127435889-127435890 on Build GRCh38
rs755252498 Chr.9: 127435904-127435904 on Build GRCh38
rs201410964 Chr.9: 127435967-127435967 on Build GRCh38
rs368448812 Chr.9: 127435896-127435896 on Build GRCh38
rs775128456 Chr.9: 127435960-127435960 on Build GRCh38
rs1026263858 Chr.9: 127435946-127435946 on Build GRCh38
rs762696497 Chr.9: 127435866-127435866 on Build GRCh38
rs751456508 Chr.9: 127435875-127435875 on Build GRCh38
rs774254577 Chr.9: 127435962-127435962 on Build GRCh38
rs748845978 Chr.9: 127436006-127436006 on Build GRCh38
rs763974160 Chr.9: 127435874-127435874 on Build GRCh38
rs754055399 Chr.9: 127435902-127435902 on Build GRCh38
rs375629022 Chr.9: 127435944-127435944 on Build GRCh38
rs371819507 Chr.9: 127435989-127435989 on Build GRCh38
rs779406091 Chr.9: 127436001-127436001 on Build GRCh38
rs768311239 Chr.9: 127436023-127436023 on Build GRCh38
rs368686424 Chr.9: 127435961-127435961 on Build GRCh38
rs748554777 Chr.9: 127435908-127435908 on Build GRCh38
rs769115244 Chr.9: 127435958-127435958 on Build GRCh38
rs764040524 Chr.9: 127435911-127435911 on Build GRCh38
rs1017821045 Chr.9: 127436010-127436010 on Build GRCh38
rs960901752 Chr.9: 127435915-127435915 on Build GRCh38
rs774951189 Chr.9: 127435857-127435857 on Build GRCh38
rs143127319 Chr.9: 127435898-127435898 on Build GRCh38
rs144556900 Chr.9: 127435994-127435994 on Build GRCh38
rs747569077 Chr.9: 127435950-127435950 on Build GRCh38
rs375751138 Chr.9: 127435887-127435887 on Build GRCh38
rs189004320 Chr.9: 127436027-127436027 on Build GRCh38
rs202189972 Chr.9: 127435899-127435899 on Build GRCh38
rs775068792 Chr.9: 127435864-127435867 on Build GRCh38

View Full Product Details