Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28150781-28151052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTAFR
Entrez Gene ID
5724
Gene Name
platelet activating factor receptor
Gene Aliases
PAFR
UniGene
Hs.77542
Gene Chromosome Location
Chr.1: 28147166-28193936 on Build GRCh38
Transcripts
NM_000952.4 NM_001164721.1 NM_001164722.2 NM_001164723.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907959 c.223G>A p.V75I Chr.1: 28150799-28150799 on Build GRCh38
5137840 c.114G>T p.W38C Chr.1: 28150908-28150908 on Build GRCh38
7165906 c.198C>T p.F66F Chr.1: 28150824-28150824 on Build GRCh38
1492064 c.151G>T p.E51* Chr.1: 28150871-28150871 on Build GRCh38
163795 c.10C>T p.H4Y Chr.1: 28151012-28151012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139373173 Chr.1: 28150917-28150917 on Build GRCh38
rs755272663 Chr.1: 28150951-28150951 on Build GRCh38
rs746947033 Chr.1: 28151015-28151015 on Build GRCh38
rs532356560 Chr.1: 28150873-28150873 on Build GRCh38
rs748377110 Chr.1: 28150843-28150843 on Build GRCh38
rs776355631 Chr.1: 28150918-28150918 on Build GRCh38
rs116552042 Chr.1: 28150940-28150940 on Build GRCh38
rs1027589464 Chr.1: 28150984-28150984 on Build GRCh38
rs761570927 Chr.1: 28150923-28150923 on Build GRCh38
rs576953829 Chr.1: 28150965-28150965 on Build GRCh38
rs763496374 Chr.1: 28150937-28150937 on Build GRCh38
rs1014846833 Chr.1: 28150902-28150902 on Build GRCh38
rs749373881 Chr.1: 28150999-28150999 on Build GRCh38
rs781553697 Chr.1: 28150961-28150961 on Build GRCh38
rs779921745 Chr.1: 28151008-28151008 on Build GRCh38
rs772835027 Chr.1: 28150916-28150916 on Build GRCh38
rs775331102 Chr.1: 28150897-28150897 on Build GRCh38
rs777875752 Chr.1: 28150997-28150997 on Build GRCh38
rs764800084 Chr.1: 28150932-28150932 on Build GRCh38
rs201541070 Chr.1: 28150982-28150982 on Build GRCh38
rs780437327 Chr.1: 28150815-28150815 on Build GRCh38
rs772030713 Chr.1: 28151005-28151005 on Build GRCh38
rs745774518 Chr.1: 28150875-28150875 on Build GRCh38
rs866954110 Chr.1: 28150838-28150838 on Build GRCh38
rs771940314 Chr.1: 28150886-28150886 on Build GRCh38
rs769811699 Chr.1: 28150858-28150858 on Build GRCh38
rs143590918 Chr.1: 28150971-28150971 on Build GRCh38
rs771531478 Chr.1: 28150912-28150912 on Build GRCh38
rs531259047 Chr.1: 28150986-28150986 on Build GRCh38
rs141858414 Chr.1: 28150836-28150836 on Build GRCh38
rs370085844 Chr.1: 28150901-28150901 on Build GRCh38
rs879188740 Chr.1: 28150811-28150811 on Build GRCh38
rs953296120 Chr.1: 28151021-28151021 on Build GRCh38
rs117376418 Chr.1: 28150938-28150938 on Build GRCh38
rs770821469 Chr.1: 28150814-28150814 on Build GRCh38
rs78853341 Chr.1: 28150817-28150817 on Build GRCh38
rs749942993 Chr.1: 28150935-28150935 on Build GRCh38
rs756356175 Chr.1: 28150981-28150981 on Build GRCh38
rs755182739 Chr.1: 28150827-28150827 on Build GRCh38

View Full Product Details