Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206585045-206585300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2D
Entrez Gene ID
1939
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
HCA56, LGTN
UniGene
Hs.497581
Gene Chromosome Location
Chr.1: 206572485-206612551 on Build GRCh38
Transcripts
XM_011509257.2 NM_182663.3 NM_182664.3 NM_182665.3

Gene Symbol
RASSF5
Entrez Gene ID
83593
Gene Name
Ras association domain family member 5
Gene Aliases
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3
UniGene
Hs.497579
Gene Chromosome Location
Chr.1: 206507530-206589284 on Build GRCh38
Transcripts
XM_011509257.2 NM_182663.3 NM_182664.3 NM_182665.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1927530 c.1033C>T p.R345C Chr.1: 206585224-206585224 on Build GRCh38
6528019 c.1073T>C p.V358A Chr.1: 206585264-206585264 on Build GRCh38
7418003 c.565C>T p.L189F Chr.1: 206585215-206585215 on Build GRCh38
6528020 c.1073T>C p.V358A Chr.1: 206585264-206585264 on Build GRCh38
3803667 c.1069T>G p.F357V Chr.1: 206585260-206585260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920934574 Chr.1: 206585091-206585091 on Build GRCh38
rs782312271 Chr.1: 206585210-206585210 on Build GRCh38
rs59679417 Chr.1: 206585161-206585161 on Build GRCh38
rs138549723 Chr.1: 206585093-206585093 on Build GRCh38
rs146761578 Chr.1: 206585247-206585247 on Build GRCh38
rs781952337 Chr.1: 206585131-206585131 on Build GRCh38
rs746023356 Chr.1: 206585195-206585195 on Build GRCh38
rs559509463 Chr.1: 206585122-206585122 on Build GRCh38
rs990398420 Chr.1: 206585111-206585111 on Build GRCh38
rs781991608 Chr.1: 206585238-206585238 on Build GRCh38
rs782213884 Chr.1: 206585225-206585225 on Build GRCh38
rs560512424 Chr.1: 206585128-206585128 on Build GRCh38
rs781797923 Chr.1: 206585176-206585176 on Build GRCh38
rs782757989 Chr.1: 206585142-206585142 on Build GRCh38
rs537393904 Chr.1: 206585081-206585081 on Build GRCh38
rs782387268 Chr.1: 206585229-206585229 on Build GRCh38
rs782650404 Chr.1: 206585208-206585208 on Build GRCh38
rs370977944 Chr.1: 206585132-206585132 on Build GRCh38
rs556937428 Chr.1: 206585085-206585085 on Build GRCh38
rs782753442 Chr.1: 206585251-206585251 on Build GRCh38
rs148610151 Chr.1: 206585198-206585198 on Build GRCh38
rs561503861 Chr.1: 206585209-206585209 on Build GRCh38
rs782715825 Chr.1: 206585169-206585169 on Build GRCh38
rs782607707 Chr.1: 206585190-206585190 on Build GRCh38
rs530192428 Chr.1: 206585246-206585246 on Build GRCh38
rs781905908 Chr.1: 206585145-206585145 on Build GRCh38
rs782292608 Chr.1: 206585242-206585242 on Build GRCh38
rs782615062 Chr.1: 206585224-206585224 on Build GRCh38
rs376377478 Chr.1: 206585130-206585130 on Build GRCh38
rs140228348 Chr.1: 206585117-206585117 on Build GRCh38
rs782057493 Chr.1: 206585250-206585250 on Build GRCh38
rs782493973 Chr.1: 206585187-206585187 on Build GRCh38
rs782558201 Chr.1: 206585200-206585200 on Build GRCh38

View Full Product Details