Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50300746-50300973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY7
Entrez Gene ID
113
Gene Name
adenylate cyclase 7
Gene Aliases
AC7
UniGene
Hs.513578
Gene Chromosome Location
Chr.16: 50246158-50318135 on Build GRCh38
Transcripts
NM_001114.4 NM_001286057.1 XM_011522835.2 XM_011522836.2 XM_011522837.2 XM_011522838.2 XM_011522839.2 XM_011522840.2 XM_011522842.2 XM_017022895.1 XM_017022896.1 XM_017022897.1 XM_017022898.1 XM_017022899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182039 c.1163G>A p.R388H Chr.16: 50300801-50300801 on Build GRCh38
5526347 c.1225G>A p.G409R Chr.16: 50300863-50300863 on Build GRCh38
435349 c.1210C>T p.R404W Chr.16: 50300848-50300848 on Build GRCh38
4523685 c.1228G>A p.V410I Chr.16: 50300866-50300866 on Build GRCh38
6320503 c.1119G>T p.V373V Chr.16: 50300757-50300757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866290590 Chr.16: 50300843-50300843 on Build GRCh38
rs200808995 Chr.16: 50300848-50300848 on Build GRCh38
rs774046376 Chr.16: 50300902-50300902 on Build GRCh38
rs953678354 Chr.16: 50300930-50300930 on Build GRCh38
rs543867974 Chr.16: 50300944-50300944 on Build GRCh38
rs766768367 Chr.16: 50300804-50300804 on Build GRCh38
rs188217442 Chr.16: 50300895-50300895 on Build GRCh38
rs773824037 Chr.16: 50300801-50300801 on Build GRCh38
rs907048113 Chr.16: 50300849-50300849 on Build GRCh38
rs867395534 Chr.16: 50300842-50300842 on Build GRCh38
rs758932458 Chr.16: 50300862-50300862 on Build GRCh38
rs374120409 Chr.16: 50300829-50300829 on Build GRCh38
rs575106713 Chr.16: 50300939-50300939 on Build GRCh38
rs1044473099 Chr.16: 50300830-50300830 on Build GRCh38
rs779821438 Chr.16: 50300891-50300891 on Build GRCh38
rs1026683717 Chr.16: 50300927-50300927 on Build GRCh38
rs1002680991 Chr.16: 50300871-50300871 on Build GRCh38
rs367628481 Chr.16: 50300881-50300881 on Build GRCh38
rs397708017 Chr.16: 50300932-50300932 on Build GRCh38
rs745430694 Chr.16: 50300880-50300880 on Build GRCh38
rs755300900 Chr.16: 50300827-50300827 on Build GRCh38
rs769421010 Chr.16: 50300889-50300889 on Build GRCh38
rs3833062 Chr.16: 50300930-50300930 on Build GRCh38
rs1017546691 Chr.16: 50300882-50300882 on Build GRCh38
rs370417008 Chr.16: 50300817-50300817 on Build GRCh38
rs138375097 Chr.16: 50300785-50300785 on Build GRCh38
rs966625433 Chr.16: 50300951-50300951 on Build GRCh38
rs182801202 Chr.16: 50300894-50300894 on Build GRCh38
rs750571359 Chr.16: 50300793-50300793 on Build GRCh38
rs984907922 Chr.16: 50300872-50300872 on Build GRCh38
rs560671117 Chr.16: 50300953-50300953 on Build GRCh38
rs763219848 Chr.16: 50300818-50300818 on Build GRCh38
rs558420152 Chr.16: 50300938-50300938 on Build GRCh38
rs748635962 Chr.16: 50300847-50300847 on Build GRCh38
rs780648752 Chr.16: 50300863-50300863 on Build GRCh38
rs774288601 Chr.16: 50300769-50300769 on Build GRCh38
rs754102907 Chr.16: 50300933-50300933 on Build GRCh38
rs1054346511 Chr.16: 50300774-50300774 on Build GRCh38
rs761680686 Chr.16: 50300784-50300784 on Build GRCh38

View Full Product Details