Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74642537-74642762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RORB
Entrez Gene ID
6096
Gene Name
RAR related orphan receptor B
Gene Aliases
NR1F2, ROR-BETA, RZR-BETA, RZRB, bA133M9.1
UniGene
Hs.494178
Gene Chromosome Location
Chr.9: 74497336-74687201 on Build GRCh38
Transcripts
NM_006914.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617644 c.445C>A p.L149M Chr.9: 74642623-74642623 on Build GRCh38
5671913 c.521C>T p.P174L Chr.9: 74642666-74642666 on Build GRCh38
4722421 c.481G>A p.G161S Chr.9: 74642659-74642659 on Build GRCh38
7387931 c.547A>G p.M183V Chr.9: 74642692-74642692 on Build GRCh38
5671912 c.488C>T p.P163L Chr.9: 74642666-74642666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146275488 Chr.9: 74642622-74642622 on Build GRCh38
rs367543552 Chr.9: 74642563-74642563 on Build GRCh38
rs368056882 Chr.9: 74642684-74642684 on Build GRCh38
rs779798482 Chr.9: 74642729-74642729 on Build GRCh38
rs138573421 Chr.9: 74642724-74642724 on Build GRCh38
rs768600001 Chr.9: 74642583-74642583 on Build GRCh38
rs151121778 Chr.9: 74642596-74642596 on Build GRCh38
rs370202250 Chr.9: 74642580-74642580 on Build GRCh38
rs768494153 Chr.9: 74642652-74642652 on Build GRCh38
rs764525958 Chr.9: 74642700-74642700 on Build GRCh38
rs746564329 Chr.9: 74642731-74642731 on Build GRCh38
rs758150843 Chr.9: 74642727-74642727 on Build GRCh38
rs763065753 Chr.9: 74642593-74642593 on Build GRCh38
rs571973540 Chr.9: 74642608-74642608 on Build GRCh38
rs747948403 Chr.9: 74642657-74642657 on Build GRCh38
rs767976733 Chr.9: 74642692-74642692 on Build GRCh38
rs138081728 Chr.9: 74642649-74642649 on Build GRCh38
rs766539793 Chr.9: 74642666-74642666 on Build GRCh38
rs149294914 Chr.9: 74642601-74642601 on Build GRCh38
rs138614785 Chr.9: 74642562-74642562 on Build GRCh38
rs746944144 Chr.9: 74642592-74642592 on Build GRCh38
rs573233356 Chr.9: 74642607-74642607 on Build GRCh38
rs761226242 Chr.9: 74642628-74642628 on Build GRCh38
rs189210068 Chr.9: 74642658-74642658 on Build GRCh38
rs746472889 Chr.9: 74642650-74642650 on Build GRCh38
rs975708949 Chr.9: 74642614-74642614 on Build GRCh38
rs756496356 Chr.9: 74642610-74642610 on Build GRCh38
rs201745846 Chr.9: 74642618-74642618 on Build GRCh38
rs751596774 Chr.9: 74642597-74642597 on Build GRCh38
rs758025910 Chr.9: 74642643-74642643 on Build GRCh38
rs199713370 Chr.9: 74642584-74642584 on Build GRCh38
rs756486376 Chr.9: 74642698-74642698 on Build GRCh38
rs1009683633 Chr.9: 74642591-74642591 on Build GRCh38
rs1040181990 Chr.9: 74642576-74642576 on Build GRCh38
rs759595738 Chr.9: 74642686-74642686 on Build GRCh38
rs144622748 Chr.9: 74642693-74642693 on Build GRCh38
rs750627186 Chr.9: 74642588-74642588 on Build GRCh38
rs774427225 Chr.9: 74642667-74642667 on Build GRCh38
rs765441867 Chr.9: 74642586-74642586 on Build GRCh38
rs745494838 Chr.9: 74642638-74642638 on Build GRCh38
rs183842881 Chr.9: 74642655-74642655 on Build GRCh38

View Full Product Details