Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92534884-92535109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM48
Entrez Gene ID
84060
Gene Name
RNA binding motif protein 48
Gene Aliases
C7orf64, HSPC304, MGC16142
UniGene
Hs.147829
Gene Chromosome Location
Chr.7: 92528709-92540481 on Build GRCh38
Transcripts
NM_032120.3 XM_005250636.4 XM_006716149.3 XM_006716150.3 XM_017012709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259862 c.1000C>T p.R334W Chr.7: 92534953-92534953 on Build GRCh38
69965 c.982A>G p.T328A Chr.7: 92534935-92534935 on Build GRCh38
79587 c.993T>C p.N331N Chr.7: 92534946-92534946 on Build GRCh38
6562987 c.961A>G p.M321V Chr.7: 92534914-92534914 on Build GRCh38
1093072 c.989C>T p.A330V Chr.7: 92534942-92534942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759839279 Chr.7: 92535029-92535029 on Build GRCh38
rs761150681 Chr.7: 92535050-92535050 on Build GRCh38
rs764476092 Chr.7: 92535055-92535055 on Build GRCh38
rs67142609 Chr.7: 92535004-92535004 on Build GRCh38
rs759306015 Chr.7: 92535032-92535035 on Build GRCh38
rs114644858 Chr.7: 92534973-92534973 on Build GRCh38
rs765029176 Chr.7: 92535059-92535060 on Build GRCh38
rs753178655 Chr.7: 92534942-92534942 on Build GRCh38
rs755376286 Chr.7: 92534939-92534939 on Build GRCh38
rs754410662 Chr.7: 92535068-92535068 on Build GRCh38
rs777288338 Chr.7: 92534945-92534945 on Build GRCh38
rs926781785 Chr.7: 92534962-92534962 on Build GRCh38
rs745581074 Chr.7: 92534984-92534984 on Build GRCh38
rs375742243 Chr.7: 92534992-92534992 on Build GRCh38
rs202023856 Chr.7: 92535066-92535067 on Build GRCh38
rs193219499 Chr.7: 92534961-92534961 on Build GRCh38
rs767882287 Chr.7: 92535038-92535038 on Build GRCh38
rs750651745 Chr.7: 92534994-92535007 on Build GRCh38
rs572555942 Chr.7: 92534915-92534915 on Build GRCh38
rs183873266 Chr.7: 92535045-92535045 on Build GRCh38
rs376528053 Chr.7: 92534919-92534919 on Build GRCh38
rs561621052 Chr.7: 92534995-92534995 on Build GRCh38
rs774655096 Chr.7: 92535025-92535025 on Build GRCh38
rs751893937 Chr.7: 92534930-92534930 on Build GRCh38
rs199804649 Chr.7: 92534943-92534943 on Build GRCh38
rs544724938 Chr.7: 92534940-92534940 on Build GRCh38
rs988141044 Chr.7: 92534920-92534920 on Build GRCh38
rs748724893 Chr.7: 92534953-92534953 on Build GRCh38
rs370711555 Chr.7: 92535075-92535075 on Build GRCh38
rs756801361 Chr.7: 92534954-92534954 on Build GRCh38
rs766619338 Chr.7: 92535024-92535024 on Build GRCh38
rs544504653 Chr.7: 92534988-92534988 on Build GRCh38
rs768504224 Chr.7: 92535001-92535001 on Build GRCh38

View Full Product Details