Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35857190-35857447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRPK1
Entrez Gene ID
6732
Gene Name
SRSF protein kinase 1
Gene Aliases
SFRSK1
UniGene
Hs.443861
Gene Chromosome Location
Chr.6: 35833034-35921180 on Build GRCh38
Transcripts
NM_003137.4 NR_034069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187127 c.1556C>T p.S519F Chr.6: 35857325-35857325 on Build GRCh38
3075714 c.1616G>A p.C539Y Chr.6: 35857265-35857265 on Build GRCh38
3626086 c.1243C>T p.L415L Chr.6: 35857314-35857314 on Build GRCh38
7283517 c.1588A>G p.T530A Chr.6: 35857293-35857293 on Build GRCh38
4864255 c.1248C>A p.I416I Chr.6: 35857309-35857309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780378379 Chr.6: 35857264-35857264 on Build GRCh38
rs528481860 Chr.6: 35857374-35857374 on Build GRCh38
rs374105061 Chr.6: 35857378-35857378 on Build GRCh38
rs990562091 Chr.6: 35857248-35857248 on Build GRCh38
rs781703276 Chr.6: 35857396-35857396 on Build GRCh38
rs753952396 Chr.6: 35857249-35857249 on Build GRCh38
rs773451762 Chr.6: 35857309-35857309 on Build GRCh38
rs757771876 Chr.6: 35857248-35857249 on Build GRCh38
rs748675506 Chr.6: 35857279-35857279 on Build GRCh38
rs747128346 Chr.6: 35857229-35857231 on Build GRCh38
rs900643673 Chr.6: 35857374-35857374 on Build GRCh38
rs373615654 Chr.6: 35857342-35857342 on Build GRCh38
rs34463636 Chr.6: 35857244-35857245 on Build GRCh38
rs61731461 Chr.6: 35857364-35857364 on Build GRCh38
rs777456269 Chr.6: 35857253-35857253 on Build GRCh38
rs746682483 Chr.6: 35857235-35857235 on Build GRCh38
rs989367949 Chr.6: 35857239-35857239 on Build GRCh38
rs772527528 Chr.6: 35857295-35857295 on Build GRCh38
rs761040725 Chr.6: 35857312-35857312 on Build GRCh38
rs375404434 Chr.6: 35857358-35857358 on Build GRCh38
rs55707453 Chr.6: 35857255-35857255 on Build GRCh38
rs35559074 Chr.6: 35857408-35857408 on Build GRCh38
rs376498696 Chr.6: 35857218-35857218 on Build GRCh38
rs373399874 Chr.6: 35857365-35857365 on Build GRCh38
rs750849022 Chr.6: 35857379-35857379 on Build GRCh38
rs756298367 Chr.6: 35857263-35857263 on Build GRCh38
rs778938649 Chr.6: 35857271-35857271 on Build GRCh38
rs778994162 Chr.6: 35857390-35857390 on Build GRCh38
rs759175583 Chr.6: 35857219-35857219 on Build GRCh38
rs182611157 Chr.6: 35857389-35857389 on Build GRCh38
rs764889990 Chr.6: 35857333-35857333 on Build GRCh38
rs749464029 Chr.6: 35857270-35857270 on Build GRCh38
rs192146058 Chr.6: 35857247-35857247 on Build GRCh38
rs759233607 Chr.6: 35857331-35857331 on Build GRCh38
rs531962877 Chr.6: 35857242-35857242 on Build GRCh38
rs564286428 Chr.6: 35857373-35857373 on Build GRCh38
rs776413587 Chr.6: 35857329-35857329 on Build GRCh38
rs764836885 Chr.6: 35857232-35857232 on Build GRCh38
rs760351696 Chr.6: 35857262-35857262 on Build GRCh38
rs771188043 Chr.6: 35857328-35857328 on Build GRCh38

View Full Product Details